Skip to Content
Merck
All Photos(3)

Key Documents

G2625

Sigma-Aldrich

Gelatin from porcine skin

gel strength ~175 g Bloom, Type A

Synonym(s):

Teleostean gelatine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.61

biological source

Porcine skin

Quality Level

type

Type A

form

powder

concentration

65-85% (biuret)

technique(s)

cell culture | mammalian: suitable

solubility

H2O: soluble 50 mg/mL

Looking for similar products? Visit Product Comparison Guide

Application

This product is recommended for use as a cell culture substratum at 1-5 μg/cm2 or 0.5-50 μg/mL. The optimal concentration does depend on cell type as well as the application and research objectives.

Gelatin has been used in many applications. It has use in coating cell culture to improve attachment of cells, being added to PCR to stabilize Taq DNA, as a blocking reagent in Western blotting, ELISA, and immunochemistry, and as a component of media for species differentiation in bacteriology. As a biocompatible polymer, it has used as a delivery vehicle for release of active biomolecules and in generation of scaffolds for tissue engineering applications. In the pharmaceutical industry, geltan can be used as a suspending and encapsulating agent, among other applications.

Components

Gelatin is a heterogeneous mixture of water-soluble proteins of high average molecular masses, present in collagen. Proteins are extracted by boiling the relevant skin, tendons, ligaments, bones, etc. in water. Type A gelatin is derived from acid-cured tissue. Type B is derived from lime-cured tissue.

Caution

Dry gelatin, when stored in airtight containers at room temperature, will remain unchanged for many years. When heated at 100°C in the presence of air, it swells becomes soft and disintegrates to a carbonaceous mass with evolution of pyridine bases and ammonia.

Preparation Note

This product is derived from porcine skin. Gelatin is soluble in hot than in cold water. It is practically insoluble in most organic solvents such as alcohol, chloroform, carbon disulfide, carbon tetrachloride, ether, benzene, acetone, and oils. The Bloom number, determined by the Bloom gelometer, is an indication of the strength of a gel formed from a solution of the known concentration. The Bloom number is proportional to the average molecular mass. Bloom numbers of porcine skin Gelatin vary from 90 to 300 g. This product has a gel strength of 175.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J W Bulte et al.
Magnetic resonance in medicine, 25(1), 148-157 (1992-05-01)
Human lymphocytes were labeled with biotinylated anti-lymphocyte-directed monoclonal antibodies, to which streptavidin and subsequently biotinylated dextran-magnetite particles were coupled. This labeling resulted in a strong and selective negative contrast enhancement of lymphocyte suspensions at 2.0 T, caused predominantly by the
Yu-Dong Ma et al.
Environmental toxicology and pharmacology, 48, 63-75 (2016-10-22)
Migration of vascular smooth muscle cells (VSMCs) contributes to intimal hyperplasia and other vascular diseases. Caveolin-1 (Cav-1) has been recognized as a proliferative inhibitor of VSMCs and is likely to be an important regulator of VSMC migration. The underlying mechanism
Souad Djediai et al.
International journal of molecular sciences, 22(23) (2021-12-11)
Epithelial-to-mesenchymal transition (EMT) recapitulates metastasis and can be induced in vitro through transforming growth factor (TGF)-β signaling. A role for MMP activity in glioblastoma multiforme has been ascribed to EMT, but the molecular crosstalk between TGF-β signaling and membrane type
François Portier et al.
Langmuir : the ACS journal of surfaces and colloids, 33(45), 12916-12925 (2017-11-01)
Collagen and its denatured form, gelatin, are biopolymers of fundamental interest in numerous fields ranging from living tissues to biomaterials, food, and cosmetics. This study aims at characterizing mixtures of those biopolymers at high concentrations (up to 100 mg·mL-1) at
Samuel Burke Nanni et al.
Biomarkers in cancer, 8, 77-87 (2016-05-27)
The low-density lipoprotein receptor-related protein 1 (LRP-1) is a multiligand endocytic receptor, which plays a pivotal role in controlling cytoskeleton dynamics during cancer cell migration. Its rapid endocytosis further allows efficient clearance of extracellular ligands. Concanavalin-A (ConA) is a lectin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service