Skip to Content
Merck
All Photos(2)

Documents

G133

Sigma-Aldrich

GR 89696 fumarate salt

solid

Synonym(s):

4-[(3,4-Dichlorophenyl)acetyl]-3-(1-pyrrolidinylmethyl)-1-piperazinecarboxylic acid methyl ester fumarate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H25Cl2N3O3 · C4H4O4
CAS Number:
Molecular Weight:
530.40
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

solid

color

off-white

solubility

ethanol: 4.0 mg/mL
DMSO: >50 mg/mL
H2O: 8.5 mg/mL

SMILES string

OC(=O)\C=C\C(O)=O.COC(=O)N1CCN(C(CN2CCCC2)C1)C(=O)Cc3ccc(Cl)c(Cl)c3

InChI

1S/C19H25Cl2N3O3.C4H4O4/c1-27-19(26)23-8-9-24(15(13-23)12-22-6-2-3-7-22)18(25)11-14-4-5-16(20)17(21)10-14;5-3(6)1-2-4(7)8/h4-5,10,15H,2-3,6-9,11-13H2,1H3;1-2H,(H,5,6)(H,7,8)/b;2-1+

InChI key

ABTNETSDXZBJTE-WLHGVMLRSA-N

Gene Information

human ... OPRK1(4986)

Application

GR 89696 fumarate salt has been used as a κ κ2 opioid receptor (KOR) agonist to study its anti-scratching and antipruritic effects on monkeys.

Biochem/physiol Actions

GR 89696 fumarate is a selective κ2 opioid receptor agonist.

Legal Information

Sold for research purposes under agreement from Glaxo­Smith­Kline

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Woong Mo Kim et al.
Anesthesia and analgesia, 113(4), 934-940 (2011-07-27)
Although bone cancer-related pain is one of the most disruptive symptoms in patients with advanced cancer, patients are often refractory to pharmacological treatments; thus, more effective treatments for bone cancer pain are needed. We evaluated the analgesic efficacy of and
Baojun Gu et al.
The Journal of urology, 171(1), 472-477 (2003-12-11)
The nonselective kappa opioid receptor agonist ethylketocyclazocine suppresses external urethral sphincter (EUS) reflexes in cats. We examined the role of spinal kappa-opioid receptor subtypes in the control of EUS function in rats using selective kappa-1 (U-50,488) or kappa-2 (GR-89,696) opiate
Peter S Talbot et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 46(3), 484-494 (2005-03-08)
Brain kappa-opioid receptors (ORs) may be involved in several pathologic conditions, such as addiction, psychosis, and seizures. (+/-)-4-Methoxycarbonyl-2-[(1-pyrrolidinylmethyl]-1-[(3,4-dichlorophenyl)acetyl]-piperidine (GR89696) is a potent and selective kappa-OR agonist. The (-)-isomer, GR103545, is the active enantiomer of GR89696. The aim of this study
Yu B Lishmanov et al.
Bulletin of experimental biology and medicine, 145(6), 696-699 (2008-12-27)
Chronic treatment with opioid receptor ligands: nonselective peptide opioid receptor agonist dalargin (intraperitoneally in a dose of 1 mg/kg), selective nonpeptide kappa-receptor agonist GR 89696 (subcutaneously in a dose of 0.03 mg/kg), nonselectrive nonpeptide antagonist quadazocine (subcutaneously in a dose
Bent W Schoultz et al.
European journal of nuclear medicine and molecular imaging, 37(6), 1174-1180 (2010-02-17)
The recent development in radiosynthesis of the (11)C-carbamate function increases the potential of [(11)C]GR103545, which for the last decade has been regarded as promising for imaging the kappa-opioid receptor (kappa-OR) with PET. In the present study, [(11)C]GR103545 was evaluated in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service