FCANOS1
Nitric Oxide Synthase Detection System, Fluorimetric
sufficient for 200 reactions
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352303
NACRES:
NA.32
Recommended Products
usage
sufficient for 200 reactions
Quality Level
availability
not available in Japan
shipped in
wet ice
storage temp.
2-8°C
Gene Information
human ... NOS1(4842) , NOS2(4843) , NOS2B(201288) , NOS2C(645740) , NOS3(4846)
Biochem/physiol Actions
Simple and specific assay for the measurement of free nitric oxide (NO) and nitric oxide synthase (NOS) activity in living cells under physiological conditions. Nitric oxide, formed from the amino acid L-arginine via the action of NOS, is a unique cell signaling molecule that functions as both an intracellular and an extracellular messenger. NO has been implicated in vasodilation, neurotransmission, cytotoxicity, and inflammation.
The standard assay for NOS activity is for solubilized cell-free enzyme preparations such as cell lysates, crude tissue extracts or purified enzymes. It measures the production of radiolabeled citrulline from radiolabeled arginine substrate (Sigma Product No. NOS-1). In contrast, the Fluorometric Cell-Associated NOS Detection System measures intracellular production of NO by a non-radiometric method. Radioisotopes are replaced by a cell-permeable diacetate derivative of 4,5-diaminofluorescein (DAF-2 DA). DAF-2 DA penetrates cells rapidly where it is hydrolyzed by intracellular esterase to DAF-2 that, in turn, reacts with NO produced by NOS to form a fluorescent triazolo-fluorescein. The fluorescent product can be quantitated using an excitation filter at 492 nm and an emission filter at 515 nm.
The standard assay for NOS activity is for solubilized cell-free enzyme preparations such as cell lysates, crude tissue extracts or purified enzymes. It measures the production of radiolabeled citrulline from radiolabeled arginine substrate (Sigma Product No. NOS-1). In contrast, the Fluorometric Cell-Associated NOS Detection System measures intracellular production of NO by a non-radiometric method. Radioisotopes are replaced by a cell-permeable diacetate derivative of 4,5-diaminofluorescein (DAF-2 DA). DAF-2 DA penetrates cells rapidly where it is hydrolyzed by intracellular esterase to DAF-2 that, in turn, reacts with NO produced by NOS to form a fluorescent triazolo-fluorescein. The fluorescent product can be quantitated using an excitation filter at 492 nm and an emission filter at 515 nm.
Legal Information
DAF-2 DA is manufactured by Daiichi Pure Chemicals Co., LTD.
US Patent No. 5,874, 590
US Patent No. 5,874, 590
Kit Components Only
Product No.
Description
- Diphenyleneiodonium Chloride (DPI) 1 mM 200 μL
- Arginine Substrate Solution for NOS Detection, Fluorimetric 2 mL
Storage Class Code
10 - Combustible liquids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Zeming Zhang et al.
Journal of cellular physiology, 234(11), 19715-19727 (2019-04-16)
Pulmonary arterial hypertension (PAH) is characterized by pulmonary vascular remodeling of the precapillary pulmonary arteries, with excessive proliferation of vascular cells. This study was performed to examine the effects of long noncoding RNA CPS1 intronic transcript 1 (CPS1-IT) on PAH
Sonia Agrawal et al.
Scientific reports, 11(1), 19813-19813 (2021-10-08)
The development of the latent phenotype of Mycobacterium tuberculosis (Mtb) in the human lungs is the major hurdle to eradicate Tuberculosis. We recently reported that exposure to nitrite (10 mM) for six days under in vitro aerobic conditions completely transforms the
D S Bredt et al.
Proceedings of the National Academy of Sciences of the United States of America, 86(22), 9030-9033 (1989-11-01)
Nitric oxide, which mediates influences of numerous neurotransmitters and modulators on vascular smooth muscle and leukocytes, can be formed in the brain from arginine by an enzymatic activity that stoichiometrically generates citrulline. We show that glutamate and related amino acids
D J Stuehr
Annual review of pharmacology and toxicology, 37, 339-359 (1997-01-01)
Research on the biological roles of nitric oxide has revealed that it functions as an important signal and effector molecule in a variety of physiologic and pathologic settings. In animals, nitric oxide is synthesized enzymatically from L-arginine through the actions
D A Geller et al.
Cancer metastasis reviews, 17(1), 7-23 (1998-04-17)
Nitric oxide (NO) is a potent biologic mediator with diverse physiologic and pathophysiologic roles. NO is produced from L-arginine by the family of nitric oxide synthase (NOS) enzymes, forming the free radical NO and citrulline as byproduct. Three distinct isoforms
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service