Skip to Content
Merck
All Photos(1)

Key Documents

EMU074451

Sigma-Aldrich

MISSION® esiRNA

targeting mouse Mapk8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCATCATGAGCAGAAGCAAACGTGACAACAATTTTTATAGTGTAGAGATTGGAGATTCTACATTCACAGTCCTAAAACGATACCAGAATTTAAAGCCTATAGGCTCAGGAGCTCAAGGAATAGTGTGTGCAGCTTATGATGCCATTCTTGAAAGAAATGTTGCAATCAAGAAGCTCAGCCGGCCATTTCAGAATCAGACCCATGCTAAGCGCGCCTACCGAGAACTAGTTCTTATGAAGTGTGTTAATCACAAAAATATAATTGGCCTTTTGAATGTTTTCACACCACAGAAATCCCTAGAAGAATTTCAAGATGTTTACATAGTCATGGAGCTCATGGATGCAAATCTTTGCCAAGTGATTCAGATGGAGTTAGATCATGAAAGAATGTCCTACCTTCTCTATCAAATGCTGTGTGGAATCAAGCACCTTCACTCTGCTG

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xu Qin et al.
Molecular carcinogenesis, 53(7), 526-536 (2013-01-30)
The c-Jun NH2 -terminal kinase (JNK) signal pathway has been implicated in the growth, cellular proliferation, and apoptosis in many kinds of carcinomas. However, the role of JNK in the development of esophageal squamous cell carcinomas (ESCCs) is unknown. To
Shantel Olivares et al.
PloS one, 9(7), e103828-e103828 (2014-08-01)
Endoplasmic reticulum (ER) stress is induced in many forms of chronic liver disease and may promote the development of hepatocellular carcinoma. The activator protein 1 (AP-1) complex is a transcription factor that promotes hepatic carcinogenesis in response to cellular stress.
Wen-Pin Cheng et al.
PloS one, 10(4), e0123235-e0123235 (2015-04-22)
The expression of TRB3 (tribbles 3), an apoptosis regulated gene, increases during endoplasmic reticulum (ER) stress. How mechanical stress affects the regulation of TRB3 in cardiomyocytes during apoptosis is not fully understood. An in vivo model of aorta-caval shunt in
Wen-Tsong Hsieh et al.
Journal of neuro-oncology, 118(2), 257-269 (2014-04-24)
Glioblastoma multiforme (GBM) is the most common and lethal type of primary brain tumor characterized by its rapid infiltration to surrounding tissues during the early stages. The fast spreading of GBM obscures the initiation of the tumor mass making the
S Goda et al.
International endodontic journal, 48(12), 1122-1128 (2014-11-14)
To investigate the effects of the c-Jun N-terminal kinase (JNK1/2) on the inflammation cytokine tumour necrosis factor-alpha (TNF-α)-enhanced production of matrix metalloproteinase-3 (MMP-3) in human dental pulp fibroblast-like cells (HPFs). HPFs were grown from pulp explants from healthy donors. Primary

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service