Skip to Content
Merck
All Photos(1)

Documents

E8409

Sigma-Aldrich

EF-24

≥98% (HPLC)

Synonym(s):

(3E,5E)-3,5-bis[(2-fluorophenyl)methylene]-4-piperidinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H15F2NO
CAS Number:
Molecular Weight:
311.33
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

yellow

solubility

DMSO: ≥10 mg/mL

storage temp.

−20°C

SMILES string

FC1=C(/C=C2CNC/C(C\2=O)=C\C3=CC=CC=C3F)C=CC=C1

InChI

1S/C19H15F2NO/c20-17-7-3-1-5-13(17)9-15-11-22-12-16(19(15)23)10-14-6-2-4-8-18(14)21/h1-10,22H,11-12H2/b15-9+,16-10+

InChI key

NIVYQYSNRUIFIF-KAVGSWPWSA-N

Application

EF-24 has been used to investigate the physiological effects and molecular targets of curcumin in Dictyostelium discoideum. It has also been used to determine the effects of ethanol extract from Zingiber zerumbet (L.) Smith rhizome (EEZZR) on diabetic retinopathy (DR) in streptozotocin (STZ) diabetic rats.

Biochem/physiol Actions

EF-24 is a IKK inhibitor and curcumin analog. EF-24 is more potent and bioavailable than curcumin, with 10-fold greater potency in cell death induction. It is also more efficacious in anti-cancer screens and less toxic than Cisplatin. EF-24 inhibits HIF-1alpha posttranscriptional activity and induces microtubule stabilization, in contrast to Curcumin. EF-24 up-regulates PTEN expression via inhibition of ubiquitination. EF-24 induces cell death through direct inhibition of IkappaB kinase (IKK), resulting in suppression in NFκB activity.
EF-24 is a IKK inhibitor and curcumin analog. EF-24 is more potent and bioavailable than curcumin, with 10-fold greater potency in cell death induction. It is also more efficacious in anti-cancer screens and less toxic than Cisplatin. EF-24 inhibits HIF-1α posttranscriptional activity and induces microtubule stabilization, in contrast to Curcumin. EF-24 up-regulates PTEN expression via inhibition of ubiquitination. EF-24 induces cell death through direct inhibition of Iκ B kinase (IKK), resulting in suppression in NFκB activity. EF-24/(3E,5E)-3,5-bis[(2-fluorophenyl)methylene]-4-piperidinone is a curcuminoid, that helps to decrease cisplatin-mediated reactive oxygen species in zebrafish. It is highly bioavailable than curcumin and has the ability to target cancer.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ebru Alp et al.
Journal of cancer research and therapeutics, 13(2), 346-350 (2017-06-24)
Cancer cells exert differential responses to chemotherapeutics and inhibitors. To the best of our knowledge, a few or no research has been performed until now to determine the effect of EF-24 and RAD001 on MDA-MB-231 breast cancer cells with regard
The ethanol extract of Zingiber zerumbet rhizomes mitigates vascular lesions in the diabetic retina
Hong T Y, et al.
Vascular Pharmacology, 76(1), 18-27 (2016)
Alice G Vassiliou et al.
Critical care (London, England), 17(5), R199-R199 (2013-09-14)
Gene expression profiling was performed via DNA microarrays in leukocytes from critically ill trauma patients nonseptic upon admission to the ICU, who subsequently developed either sepsis (n = 2) or severe sepsis and acute respiratory distress syndrome (n = 3).
Curcumin and derivatives function through protein phosphatase 2A and presenilin orthologues in Dictyostelium discoideum
Cocorocchio M, et al.
Disease models & mechanisms, 11(1), dmm032375-dmm032375 (2018)
The curcuminoid, EF-24, reduces cisplatin-mediated reactive oxygen species in zebrafish inner ear auditory and vestibular tissues
Monroe J D, et al.
Journal of Clinical Neuroscience : Official Journal of the Neurosurgical Society of Australasia, 57, 152-156 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service