Skip to Content
Merck
All Photos(1)

Key Documents

E7144

Sigma-Aldrich

Exendin-4

≥97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C184H282N50O60S
CAS Number:
Molecular Weight:
4186.57
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.32

Quality Level

Assay

≥97%

storage temp.

−20°C

SMILES string

S(CC[C@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)CNC(=O)[C@@H](NC(=O)CNC(=O)[C@@H](N)Cc9[nH]cnc9)CCC(=O)O)[C@H](O)C)Cc8ccccc8)[C@H](O)C)CO)CC(=O)O)CC(C)C)CO)CCCCN)CCC(=O)N

InChI

1S/C184H282N50O60S/c1-16-94(10)147(178(289)213-114(52-58-144(257)258)163(274)218-121(73-101-77-195-105-39-24-23-38-103(101)105)168(279)215-116(68-90(2)3)165(276)205-107(41-26-28-61-186)158(269)219-122(75-134(189)243)154(265)198-79-135(244)196-83-139(248)231-63-30-43-129(231)175(286)225-127(87-238)174(285)223-125(85-236)155(266)200-80-136(245)202-96(12)181(292)233-65-32-45-131(233)183(294)234-66-33-46-132(234)182(293)232-64-31-44-130(232)176(287)222-124(84-235)150(190)261)229-170(281)119(71-99-34-19-17-20-35-99)217-166(277)117(69-91(4)5)214-159(270)108(42-29-62-194-184(191)192)212-177(288)146(93(8)9)228-151(262)95(11)203-156(267)111(49-55-141(251)252)208-161(272)112(50-56-142(253)254)209-162(273)113(51-57-143(255)256)210-164(275)115(59-67-295-15)211-160(271)110(47-53-133(188)242)207-157(268)106(40-25-27-60-185)206-172(283)126(86-237)224-167(278)118(70-92(6)7)216-169(280)123(76-145(259)260)220-173(284)128(88-239)226-180(291)149(98(14)241)230-171(282)120(72-100-36-21-18-22-37-100)221-179(290)148(97(13)240)227-138(247)82-199-153(264)109(48-54-140(249)250)204-137(246)81-197-152(263)104(187)74-102-78-193-89-201-102/h17-24,34-39,77-78,89-98,104,106-132,146-149,195,235-241H,16,25-33,40-76,79-88,185-187H2,1-15H3,(H2,188,242)(H2,189,243)(H2,190,261)(H,193,201)(H,196,244)(H,197,263)(H,198,265)(H,199,264)(H,200,266)(H,202,245)(H,203,267)(H,204,246)(H,205,276)(H,206,283)(H,207,268)(H,208,272)(H,209,273)(H,210,275)(H,211,271)(H,212,288)(H,213,289)(H,214,270)(H,215,279)(H,216,280)(H,217,277)(H,218,274)(H,219,269)(H,220,284)(H,221,290)(H,222,287)(H,223,285)(H,224,278)(H,225,286)(H,226,291)(H,227,247)(H,228,262)(H,229,281)(H,230,282)(H,249,250)(H,251,252)(H,253,254)(H,255,256)(H,257,258)(H,259,260)(H4,191,192,194)/t94-,95-,96-,97+,98+,104-,106-,107-,108-,109-,110-,111-,112-,113-,114-,115-,116-,117-,118-,119-,120-,121-,122-,123-,124-,125-,126-,127-,128-,129-,130-,131-,132-,146-,147-,148-,149-/m0/s1

InChI key

HTQBXNHDCUEHJF-XWLPCZSASA-N

Gene Information

human ... GLP1R(2740)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

His-Gly-Glu-Gly-Thr-Phe-Thr-Ser-Asp-Leu-Ser-Lys-Gln-Met-Glu-Glu-Glu-Ala-Val-Arg-Leu-Phe-Ile-Glu-Trp-Leu-Lys-Asn-Gly-Gly-Pro-Ser-Ser-Gly-Ala-Pro-Pro-Pro-Ser-NH2

General description

Exendin-4 is an incretin mimetic peptide, which is composed of 39 amino acids. It is an analog of glucagon-like peptide 1 (GLP-1), and an insulinotropic agent, with a long half-life. Exendin-4 was historically isolated from the venom of Gila monster lizard called Heloderma suspectum.

Application

Exendin-4 has been used as incretin mimetics for the treatment in HepG2 cells to exclude the influence of auxiliary material. It has also been used as an r (GLP-1R) agonist to exclude interference from auxiliary material.
Exendin-4 has been used for the treatment of ob/ob mouse model to determine the glucose and insulin concentrations and homeostasis model of assessment (HOMA), and for the determination of nuclear factor-κΒ (NF-κB) p65 level in macrophages treated with exendin-4.

Biochem/physiol Actions

Activates GLP-1 (glucagon-like peptide-1) receptors to increase intracellular cAMP in pancreatic acinar cells; has no effect on VIP receptors.
Exendin-4 mimics the activity of mammalian incretin hormone glucagon-like peptide 1 (GLP-1), and thus, functions in the control of glucose. This includes the secretion of insulin in a glucose dependent manner, negative regulation of high glucagon secretion, and increased duration of stomach emptying. In type 2 diabetes patients, this peptide can be administered subcutaneously for glycemic control, when metformin is unable to produce adequate results. It promotes the neogeneration and proliferation of β-cells, and thus aids in the regeneration of pancreas. It acts as a ligand to exendin receptor, and leads to an elevation of acinar cell cAMP levels.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Synthetically produced
Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Junling Liu et al.
EBioMedicine, 41, 73-84 (2019-03-05)
Glucagon-like peptide-1 (GLP-1) and its based agents improve glycemic control. Although their attenuating effect on hepatic glucose output has drawn our attention for decades, the potential mechanisms remain unclear. Cytokine array kit was used to assess cytokine profiles in db/db
GLP-1 receptor agonist promotes brown remodelling in mouse white adipose tissue through SIRT1
Xu F, et al.
Diabetologia, 59(5), 1059-1069 (2016)
C Montrose-Rafizadeh et al.
The Journal of biological chemistry, 272(34), 21201-21206 (1997-08-22)
GLP-1-(7-36)-amide and exendin-4-(1-39) are glucagon-like peptide-1 (GLP-1) receptor agonists, whereas exendin-(9-39) is the only known antagonist. To analyze the transition from agonist to antagonist and to identify the amino acid residues involved in ligand activation of the GLP-1 receptor, we
SIRT1 mediates the effect of GLP-1 receptor agonist exenatide on ameliorating hepatic steatosis
Xu F, et al.
Diabetes, 63(11), 3637-3646 (2014)
Anujith Kumar et al.
PloS one, 8(5), e63491-e63491 (2013-05-15)
β-cell replacement may efficiently cure type 1 diabetic (T1D) patients whose insulin-secreting β-cells have been selectively destroyed by autoantigen-reactive T cells. To generate insulin-secreting cells we used two cell sources: rat multipotent adult progenitor cells (rMAPC) and the highly similar

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service