Skip to Content
Merck
All Photos(1)

Key Documents

D3321

Sigma-Aldrich

Dipeptidyl Peptidase VII human

recombinant, expressed in Sf9 cells

Synonym(s):

DPP7, Quiescent cell proline dipeptidase

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
3.4.14.-
UNSPSC Code:
12352204
NACRES:
NA.54

recombinant

expressed in Sf9 cells

Quality Level

form

solution

specific activity

≥1,500 units/μg protein
≥1500 units/μg protein

mol wt

89.1 kDa

relevant disease(s)

diabetes; cardiovascular diseases

shipped in

dry ice

storage temp.

−70°C

Application

Dipeptidyl Peptidase VII (DPP7), also known as DPP2 or quiescent cell proline dipeptidase, is a post-proline cleaving aminopeptidase that is expressed in quiescent lymphocytes . DPP7 is used to study the regulation of cell quiescence . Like DPP4, DPP7 may be useful in diabetes and vascular disease research .

Biochem/physiol Actions

DPP7 is essential for maintaining lymphocytes and fibroblasts in G(0). The inhibition of DPP7 results in apoptosis, which is mediated by the induction of c-Myc and p53 . DPP7 has strong sequence homology with prolylcarboxypeptidase and is active at both acidic and neutral pH.

Physical properties

Contains amino acids 29 to end with a C-terminal His tag, MW=89.1 kDa

Unit Definition

One unit will hydrolyze 1.0 picomole of Ala-Pro-AMC per minute at pH 7.4 at 25 deg °C

Physical form

Supplied as a solution in 40 mM Tris-HCl, pH 8.0, 110 mM NaCl, 2.2 mM KCl, 220 mM imidazole, and 20% glycerol.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Repr. 1B - Skin Corr. 1C

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martin Kruse et al.
The Journal of general physiology, 140(2), 189-205 (2012-08-02)
Phosphatidylinositol 4,5-bisphosphate (PI(4,5)P(2)) regulates activities of numerous ion channels including inwardly rectifying potassium (K(ir)) channels, KCNQ, TRP, and voltage-gated calcium channels. Several studies suggest that voltage-gated potassium (K(V)) channels might be regulated by PI(4,5)P(2). Wide expression of K(V) channels in
Dipeptidyl peptidase 2 is an essential survival factor in the regulation of cell quiescence.
Mele DA, et al.
Cell Cycle, 15, 2425-2434 (2009)
Katrin Witzel et al.
Neuropharmacology, 63(8), 1389-1403 (2012-09-12)
We examined the effects of the sulfonylurea compound NS5806 on neuronal A-type channel function. Using whole-cell patch-clamp we studied the effects of NS5806 on the somatodendritic A-type current (I(SA)) in cultured hippocampal neurons and the currents mediated by Kv4.2 channels
Lin Zhu et al.
The Journal of general and applied microbiology, 58(3), 199-209 (2012-08-11)
Proteolytic degradation is one of the serious bottlenecks limiting the yields of heterologous protein production by Aspergillus oryzae. In this study, we selected a tripeptidyl peptidase gene AosedD (AO090166000084) as a candidate potentially degrading the heterologous protein, and performed localization
Mahesh Kamate et al.
Neurology India, 60(3), 316-320 (2012-07-25)
Neuronal ceroid lipofuscinosis is a group of progressive neurodegenerative disorders characterized by accumulation of ceroid lipopigment in lysosomes in neurons and other cell types. This study is a retrospective review of charts of patients with a diagnosis of infantile and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service