Skip to Content
Merck
All Photos(1)

Key Documents

CLS4681

Corning® 384 well microplate

black cyclic olefin copolymer (COC) (COC), flat bottom, sterile, lid, pack of 20

Synonym(s):

384 multiwell plates, 384 well low volume plates, 384 well microplates, 384 well microtiter plates, 384 well plates, 384 well tissue culture treated plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.15

material

black cyclic olefin copolymer (COC) (COC)
clear bottom
flat bottom

sterility

sterile

feature

lid
skirt
black cyclic olefin copolymer (COC)
half area, HCS, film bottom

packaging

pack of 20

manufacturer/tradename

Corning 4681

well volume

82 μL

wells

384 wells

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

Application

Corning® 384 well microplates are suitable for high-resolution cellular imaging microscopy.

Features and Benefits

  • Microplates made of cyclic olefin copolymer (COC) material are chemical resistant (including DMSO) and have high mechanical stability
  • Microplates with ultra-clear film with 127 μm thickness and inter- and intra-well film bottom flatness within 50 μm and 10 μm, respectively, optimized for high content applications
  • Low auto-fluorescence and birefringence

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service