Skip to Content
Merck
All Photos(1)

Key Documents

CLS3572

Corning® 384 well microplate, low flange

white polystyrene, square, non-sterile, lid: no, pack of 50

Synonym(s):

384 multiwell plates, 384 well low volume plates, 384 well microplates, 384 well microtiter plates, 384 well plates, 384 well tissue culture treated plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.15
material:
flat wells
polystyrene
square
white bottom
white polystyrene
maximum volume:
112 μL
size:
384 wells
well volume:
112 μL
well working volume:
20-80 μL
working volume:
80 μL
Pricing and availability is not currently available.

material

flat wells
polystyrene
square
white bottom
white polystyrene

sterility

non-sterile

feature

lid: no
skirt

packaging

pack of 50

manufacturer/tradename

Corning 3572

maximum volume

112 μL

size

384 wells

well volume

112 μL

well working volume

20-80 μL

working volume

80 μL

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

  • Corning® 384 well microplate, low flange is suitable for immobilization of large molecules
  • Untreated white wall plates with minimal well-to-well crosstalk during luminescent assays
  • Binding capacity of approximately 100 to 200ng IgG/cm²
  • Standard microplate footprint and dimensions
  • Medium binding polystyrene hydrophobic surface enables biomolecules binding through passive interactions
  • Ideal for sensitive assays and plate readers

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service