Skip to Content
Merck
All Photos(1)

Key Documents

PSA1

85011403, mouse embryo, Teratocarcinoma

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

PSA1, 85011403

biological source

mouse embryo

growth mode

Suspension

karyotype

n = 40

morphology

Teratocarcinoma

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse SIM embryo teratocarcinoma

Cell Line Description

Thioguanine and ouabain resistant derivative of SIM. When cultured in absence of STO feeder cells it is stimulated to develop cell aggregates which differentiate resulting in formation of layer of endodermal cells covering aggregates.

Application

Differentiation and tumourigenicity studies

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C. Requires STO feeder layer.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service