Skip to Content
Merck
All Photos(1)

Documents

C3647

Sigma-Aldrich

Z-L-Lys-SBzl hydrochloride

≥98% (TLC), suitable for ligand binding assays

Synonym(s):

N-α-Cbz-L-lysine thiobenzyl ester hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H26N2O3S · HCl
CAS Number:
Molecular Weight:
422.97
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

product name

Z-L-Lys-SBzl hydrochloride,

Assay

≥98% (TLC)

form

powder

technique(s)

ligand binding assay: suitable

color

white

storage temp.

−20°C

SMILES string

Cl.NCCCC[C@H](NC(=O)OCc1ccccc1)C(=O)SCc2ccccc2

InChI

1S/C21H26N2O3S.ClH/c22-14-8-7-13-19(20(24)27-16-18-11-5-2-6-12-18)23-21(25)26-15-17-9-3-1-4-10-17;/h1-6,9-12,19H,7-8,13-16,22H2,(H,23,25);1H/t19-;/m0./s1

InChI key

PNDSXGXPDJRQAV-FYZYNONXSA-N

Application

Z-L-Lys-SBzl hydrochloride has been used as a thioesther substrate in photometric assay and as substrate in determining mast cell protease enzyme activity.

Biochem/physiol Actions

N-α-Cbz-L-lysine thiobenzyl ester (Z-L-Lys-SBzl) is used to identify and characterize specific protease/esterase/granzyme activities associated with lymphocyte cytoplasmic granules.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

The major component of Na-CBZ-L-lysine thiobenzyl ester (BLT)-specific proteases in cytoplasmic granules of murine intraepithelial lymphocytes is granzyme A.
Kato Y, Yokochi T, Sasaki K, et al.
Immunobiol., 196, 465-474 (1997)
The isolation and purification of a dual specific mast cell-derived protease from parasitised caprine jejunal tissue
Macaldowie CN, et al.
Research in Veterinary Science, 64(1), 17-24 (1998)
B A Williams et al.
Experimental cell research, 233(1), 187-197 (1997-05-25)
The effect of 2-chloroadenosine (2CA), an adenosine receptor agonist, on the activation status of mouse natural killer (NK) cells was determined. Splenic lymphocytes incubated with 2CA exocytosed an NK cell-associated granzyme with N alpha-CBZ-L-lysine thiobenzyl ester (BLT) esterase activity in
Andrea Kocsis et al.
Journal of immunology (Baltimore, Md. : 1950), 185(7), 4169-4178 (2010-09-08)
The complement system, an essential part of the innate immune system, can be activated through three distinct routes: the classical, the alternative, and the lectin pathways. The contribution of individual activation pathways to different biological processes can be assessed by
Gregory D Rak et al.
PLoS biology, 9(9), e1001151-e1001151 (2011-09-21)
Accumulation of filamentous actin (F-actin) at the immunological synapse (IS) is a prerequisite for the cytotoxic function of natural killer (NK) cells. Subsequent to reorganization of the actin network, lytic granules polarize to the IS where their contents are secreted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service