Skip to Content
Merck
All Photos(4)

Documents

C3394

Sigma-Aldrich

Cordycepin

from Cordyceps militaris, ≥98% (HPLC), powder, adenosine analogue

Synonym(s):

3′-Deoxyadenosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13N5O3
CAS Number:
Molecular Weight:
251.24
Beilstein:
35194
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

Cordycepin, from Cordyceps militaris

biological source

Cordyceps militaris

Quality Level

form

powder

antibiotic activity spectrum

fungi

Mode of action

DNA synthesis | interferes
enzyme | inhibits

storage temp.

−20°C

SMILES string

Nc1ncnc2n(cnc12)[C@@H]3O[C@H](CO)C[C@H]3O

InChI

1S/C10H13N5O3/c11-8-7-9(13-3-12-8)15(4-14-7)10-6(17)1-5(2-16)18-10/h3-6,10,16-17H,1-2H2,(H2,11,12,13)/t5-,6+,10+/m0/s1

InChI key

OFEZSBMBBKLLBJ-BAJZRUMYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: nucleoside

Biochem/physiol Actions

Cordycepin is an adenosine analogue that is readily converted to cordycepin 5′-triphosphate; can be used for 3′-end labeling of RNA.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide and Gene Regulation research. Discover more featured Cyclic Nucleotide and Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Adenylyl cyclases page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E N Kodama et al.
Biochemical pharmacology, 59(3), 273-281 (1999-12-28)
The nucleoside analogue cordycepin (3'-deoxyadenosine, 3'-dA) is substantially more cytotoxic to terminal deoxynucleotidyl transferase positive (TdT+) leukemic cells than to TdT leukemic cells in vitro in the presence of an adenosine deaminase inhibitor, deoxycoformycin (dCF), and has been considered as
Wei Su et al.
RNA (New York, N.Y.), 19(1), 1-16 (2012-11-29)
Metazoan replication-dependent histone mRNAs are only present in S-phase, due partly to changes in their stability. These mRNAs end in a unique stem-loop (SL) that is required for both translation and cell-cycle regulation. Previous studies showed that histone mRNA degradation
Thiyagarajan Ramesh et al.
Experimental gerontology, 47(12), 979-987 (2012-09-25)
Free radical-induced oxidative damage is considered to be the most important consequence of the aging process. The activities and capacities of antioxidant systems of cells decline with increased age, leading to the gradual loss of pro-oxidant/antioxidant balance and resulting in
Tianran Jia et al.
Bio-protocol, 10(14), e3680-e3680 (2021-03-05)
Steady-state mRNA levels are determined by both the rates of transcription and degradation. Regulation of mRNA stability and/or degradation are key factors that can significantly affect mRNA levels and its biological functions. mRNA stability can be measured indirectly after transcription
Guifen Wu et al.
Cell reports, 30(7), 2387-2401 (2020-02-23)
Degradation of transcripts in human nuclei is primarily facilitated by the RNA exosome. To obtain substrate specificity, the exosome is aided by adaptors; in the nucleoplasm, those adaptors are the nuclear exosome-targeting (NEXT) complex and the poly(A) (pA) exosome-targeting (PAXT) connection.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service