Skip to Content
Merck
All Photos(1)

Key Documents

ABK0014

Sigma-Aldrich

LuceBridge ELISA Kit from rabbit serum

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

antibody form

purified immunoglobulin

species reactivity

human (IgG)

shipped in

wet ice

storage temp.

2-8°C

General description

A Protein Conformational Array (PCA) in a sandwich ELISA format where the plate is coated with a panel of antibodies raised against peptides derived from the full length protein sequence of Ranibizumab. Taken individually, each of these antibodies is strongly antigenic to the peptide sequence that was used in its production. However, when these peptides are incorporated into a full length correctly folded protein, the antigenicity of many of them is masked by the three dimensional structure of the protein and only a limited number of the antibodies respond. The result is a histogram which can be likened to a ‘fingerprint′ for correctly folded Ranibizumab. For a Lucentis Biosimilar, if the protein is correctly folded and glycosylated, the ‘fingerprint′ will match that of Ranibizumab. If it is not correctly folded, previously masked peptide sequences will be exposed and will be recognized by the antibody made to that exposed sequence. In this way, changes in the ‘fingerprint′ generated by the ELISA will point out differences between the Biosimilar and authentic Ranibizumab.

Application

For research use only. Not for use in diagnostic procedures.

Other Notes

ELISA kit contains 2 96-well microplates and all the components necessary to complete an ELISA test.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xing Wang et al.
Frontiers in pharmacology, 4, 103-103 (2013-08-24)
Antibody arrays were developed to probe a monoclonal antibody's three-dimensional structure (3-D structure). Peptides with overlapping regions were designed to cover the whole mAb light chain and heavy chain, respectively, and used to generate polyclonal antibodies after the conjugation of
Yuanli Song et al.
mAbs, 10(3), 397-405 (2018-01-10)
The elucidation of antibody higher order structure (HOS) is critical in therapeutic antibody development. Since HOS determines the protein bioactivity and chemo-physical properties, this knowledge can help to ensure that the safety and efficacy attributes are not compromised. Protein conformational
Biosimilar mAb in-process Sample Higher Order Structure Analysis with Protein Conformational Array ELISA.
Michael Davies, Gan Wang, Jian Gong, Guofeng Fu and Xing Wang
Pharmaceut. Res., 1-11 (2016)
Soon Kwan Jung et al.
mAbs, 6(5), 1163-1177 (2014-12-18)
Remsima (infliximab) was recently approved as the world's first biosimilar monoclonal antibody (mAb) in both the European Union and Korea. To achieve this, extensive physicochemical characterization of Remsima in relation to Remicade was conducted in order to demonstrate the highly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service