Skip to Content
Merck
All Photos(1)

Key Documents

A1559

Sigma-Aldrich

Monoclonal Anti-Biotin–Agarose antibody produced in mouse

clone BN-34, purified immunoglobulin, PBS suspension

Synonym(s):

Monoclonal Anti-Biotin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

biological source

mouse

recombinant

expressed in mouse cell line

conjugate

agarose conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

BN-34, monoclonal

form

PBS suspension

extent of labeling

2 mg antibody per mL resin

technique(s)

ELISA: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1

capacity

0.15-0.3 μmol/mL, agarose binding capacity (biotin-ligand conjugate)

application(s)

research pathology

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Biotin is an essential vitamin required for cells in living organisms and in cultures. The high binding affinity of biotin to egg white avidin or bacteria-derived streptavidin has been used to design immunoassays for detecting antigen-antibody interactions. The use of monoclonal anti-biotin IgG can enhance the sensitivity of such avidin-biotin immunoassays. The monoclonal antibody functions by selectively enlarging and bridging the avidin-biotin-enzyme complexes, thereby increasing the signals from substrate conversion. Furthermore, this antibody can be used in many other applications where biotin may be introduced as target label.
The antibody recognizes free biotin and biocytin using competitive ELISA. Using indirect ELISA or immuno­histo­chemical staining, the antibody also recognizes biotin conjugated to various immunoglobulins. Specificity is evaluated using biotinylated goat antibodies to human and rabbit antigens coated on multiwell plates.

Immunogen

Biotin-KLH conjugate

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Chromatin immunoprecipitation (1 paper)
Monoclonal Anti-Biotin-Agarose antibody produced in mouse has been used in:
  • pulldown assay
  • coimmunoprecipitation 
  • chromatin immunoprecipitation
  • purification of biotinylated compounds and biotin-containing compounds
Mouse monoclonal anti-biotin antibody was used to identify by western blot S-nitrosylated proteins that had been labeled with a biotin moeity based on the biotin-switch method. No reactivity was visible on the blots when the biotin switch method was inhibited by treatment with 100 mM dithiothreitol showing that this antibody shows no unspecific cross reactivity. The mouse monoclonal anti-biotin antibody does cross react with Arabidopsis plant by western blot, no cross reactivity with unbiotinylated proteins.
In some applications, localization of biotinylated probes with avidin produces high background levels. Anti-biotin reagents may be substituted for avidin to decrease non-specific binding.
The antibody may be used in amplification techniques, ELISA, immunoblotting, in situ nucleic acid hybridization, flow cytometry, image analysis or confocal microscopy.
In some applications, localization of biotinylated probes with avidin produces high background levels. Anti-biotin reagents may be substituted for avidin to decrease non-specific binding.

Physical form

Suspension in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide

Preparation Note

Prepared using purified antibody coupled to cyanogen bromide activated agarose

Other Notes

Polyclonal anti-biotin antibodies were shown to react with in vivo biotinylated proteins but not monoclonal anti-biotin antibodies.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Namrata D Udeshi et al.
Nature methods, 14(12), 1167-1170 (2017-10-19)
Although purification of biotinylated molecules is highly efficient, identifying specific sites of biotinylation remains challenging. We show that anti-biotin antibodies enable unprecedented enrichment of biotinylated peptides from complex peptide mixtures. Live-cell proximity labeling using APEX peroxidase followed by anti-biotin enrichment
A two-step strategy to visually identify molecularly imprinted polymers for tagged proteins
Brandis A, et al.
Journal of Separation Science, 40(16), 3358-3367 (2017)
Protein phosphatase 1c associated with the cardiac sodium calcium exchanger 1 regulates its activity by dephosphorylating serine 68-phosphorylated phospholemman
Hafver T, et al.
The Journal of biological chemistry, 291(9), 4561-4579 (2016)
Transient dsDNA breaks during pre-replication complex assembly
Rampakakis E and Zannis-Hadjopoulos M
Nucleic Acids Research, 37(17), 5714-5724 (2009)
Alexander Brandis et al.
Journal of separation science, 40(16), 3358-3367 (2017-06-13)
A practical and relatively simple method to identify molecularly imprinted polymers capable of binding proteins via the molecular tagging (epitope-like) approach has been developed. In our two-step method, we first challenge a previously obtained anti-tag molecularly imprinted polymer with a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service