Skip to Content
Merck
All Photos(2)

Key Documents

408-05A

Sigma-Aldrich

Human Fibroblast-Like Synoviocytes: HFLS, adult

Synonym(s):

HFLS cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.81

biological source

human synovial tissues (normal)

Quality Level

packaging

pkg of 500,000 cells

manufacturer/tradename

Cell Applications, Inc

growth mode

Adherent

karyotype

2n = 46

morphology

Fibroblast-like

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

arthritis

shipped in

dry ice

storage temp.

−196°C

General description

Lot specific orders are not able to be placed through the web. Contact your local sales rep for more details.

Normal HFLS provide an excellent cellular model for studying the normal and pathological physiology of synoviocytes and development of joint diseases.

Normal HFLS have been used in numerous research studies to:
  • Study changes in gene expression in synoviocytes stimulated with TNFα (Suggiyama, 2002)
  • Demonstrate that TNF-like weak inducer of apoptosis (TWEAK) contributes to joint inflammation by inducing chemokines such as MIP-1β (CCL-4), lymphotactin (XCL-1), IFN-γ-inducible protein 10 (IP-10) (CXCL-10), MCP-1 (CCL-2), and RANTES (CCL-5), as well as the matrix metalloprotease-9, suggesting TWEAK as a new therapeutic target (Perper, 2006)
  • Demonstrate the role of miR-124a in arthritis pathogenesis (Nakamachi, 2009)
  • Show that C/EBPβ regulates expression of metalloproteinases and ADAMTS family members in synoviocytes stimulated with IL-1β (Tsushima, 2012); and that HMW-HA suppressed ADAMTS4 mRNA and protein expression via CD44, p38 MAPK and JNK pathways (Kataoka, 2013)
  • Evaluate the anti-inflammatory and antirheumatic activity of various compounds, such as NF-κB inhibitors (Wen, 2006), AGIX-4207 (Kunsch, 2005), HA–methotrexate conjugates (Homma, 2009) and bucillamine (Oki, 2009)
  • Show that reactive arthritis triggered by chlamydial infection is mediated by TLR2 induced IL-6 production in synoviocytes (Konomi, 2009); and investigate the mechanisms of arthritis-like syndrome in patients infected with chikungunya (CHIK) virus (Xu, 2013)
  • Suggest that only leukocyte-poor, RBC-free platelet-rich plasma should be used in clinical orthopaedics because leukocytes and RBCs cause synoviocyte death and proinflammatory mediator production (Brown, 2014)
  • Identify that cartilage link protein and MAGP2 can be used as specific markers to distinguish chondrocytes and synovial cells (Rapko, 2007, 2010)
  • Create, along with human dermal fibroblasts, induced pluripotent stem cells (iPSC) by using the now classic “Yamanaka cocktail”, the discovery for which Dr. Shinya Yamanaka was awarded the Nobel Prize in 2012 (Takahashi, 2007)

Additionally, in parallel with HFLS isolated from joints of patients with rheumatoid arthritis, normal HFLS were used to:
  • Identify causing agents (such as uric acid crystals or platelet microparticles) and study the immunopathological mechanisms and signal transduction pathways leading to joint inflammation in rheumatoid arthritis (Chen, 2011a; Hsu, 2012; Mathieu, 2008; Tsuji, 2012), and to demonstrate the role of estrogen signaling in increasing inflammation (Galal, 2008)
  • Investigate anti-inflammatory properties of herbal compound Sinomenine suggested for rheumatoid arthritis treatment (Chen, 2011b)
  • Study the effects of extracellular matrix composition on cell attachment and migration relevant to T-cell function in inflamed tissues (Evanko, 2012)

Finally, all three types of HFLS (normal, RA and OA) were used to investigate the role of human endogenous retroviruses (HERVs) in development of rheumatoid arthritis, and suggest that activated expression of different forms of HERV contribute to development of rheumatoid arthritis symptoms by different mechanisms (Freimanis, 2010).

Cell Line Origin

Bone

Application

synoviocyte physiology, joint homeostasis, production of collagen, glycoproteins, lubrican, vimentin and hyaluronic acid, gene expression, signal transduction pathways, enzyme production, evaluation of anti-inflammatory agents, effects of extracellular matrix, cell attachment, migration, integrin study, proinflammatory mediator production

Components

Basal Medium containing 10% FBS & 10% DMSO

Preparation Note

  • 2nd passage, >500,000 cells in Basal Medium containing 10% FBS & 10% DMSO
  • Can be cultured at least 5 doublings

Subculture Routine

Please refer to the HFLS Culture Protocol.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service