Skip to Content
Merck
All Photos(1)

Documents

264M-9

Sigma-Aldrich

Hepatocyte Specific Antigen (Hep Par-1) (OCH1E5) Mouse Monoclonal Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

OCH1E5, monoclonal

description

For In Vitro Diagnostic Use in Select Regions (See Chart)

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (264M-94)
vial of 0.5 mL concentrate (264M-95)
bottle of 1.0 mL predilute (264M-97)
vial of 1.0 mL concentrate (264M-96)
bottle of 7.0 mL predilute (264M-98)

manufacturer/tradename

Cell Marque

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:100-1:500

isotype

IgG1κ

control

liver

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

General description

Anti-hepatocyte specific antigen, also known as anti-Hep-Par1 (hepatocyte paraffin 1),recognizes both benign and malignant liver-derived tissues including such tumors ashepatocellular carcinoma. It recognizes both normal adult and fetal liver tissue. The typicalpattern is granular cytoplasmic staining. This antibody is useful in identifying hepatocellularcarcinomas.

Quality


IVD

IVD

IVD

RUO

Linkage

Hepatocyte Specific Antigen (Hep-Par1)(OCH1E5) Positive Control Slides, Product No. 264S, are available for immunohistochemistry (formalin-fixed, paraffin-embedded sections).

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide

Preparation Note

Download the IFU specific to your product lot and formatNote: This requires a keycode which can be found on your packaging or product label.

Other Notes

For Technical Service please contact: 800-665-7284 or email: service@cellmarque.com

Legal Information

Cell Marque is a trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W M Tsui et al.
The American journal of surgical pathology, 23(1), 34-48 (1999-01-15)
Hepatic angiomyolipoma (AML) is frequently misdiagnosed. HMB-45 is a promising immunomarker for this tumor that leads to recognition of some AMLs with unusual morphology. The purpose of this collaborative study is to better define the morphologic variations of AML. Thirty
A Maitra et al.
American journal of clinical pathology, 115(5), 689-694 (2001-05-11)
Hepatocyte paraffin 1 (Hep Par 1) is a monoclonal antibody considered almost specific for normal and neoplastic hepatocytes, that can be used on formalin-fixed paraffin-embedded tissues. Hep Par 1 reactivity has been demonstrated consistently in hepatocellular carcinomas and hepatoblastomas but
Tad J Wieczorek et al.
American journal of clinical pathology, 118(6), 911-921 (2002-12-11)
We compared the effectiveness of thyroid transcription factor-1 (TTF-1, cytoplasmic reactivity) and hepatocyte antigen (HPA) as markers for characterization of hepatocellular carcinoma (HCC) and as discriminators to distinguish HCC from its histologic and cytologic mimics. Formalin-fixed, paraffin-embedded sections of 258
M Fasano et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 11(10), 934-938 (1998-10-31)
The distinction of hepatoblastoma, especially the embryonal type, from other small, round-cell tumors of childhood can sometimes be difficult. Polyclonal anticarcinoembryonic antigen (pCEA) and Hepatocyte Paraffin 1 (Hep Par 1) are immunohistochemical markers that are useful in the diagnosis of
Peiguo G Chu et al.
The American journal of surgical pathology, 26(8), 978-988 (2002-08-10)
Hepatocyte monoclonal antibody (Hep) (alternatively Hep Par 1 for Hep paraffin 1) has been reported to stain normal hepatic tissue and hepatocellular carcinoma (HCC) with high specificity. We have studied the Hepatocyte expression in 96 cases of HCC and 311

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service