Skip to Content
Merck
All Photos(1)

Documents

12023C

Sigma-Aldrich

Newborn Bovine Serum

New Zealand origin

Synonym(s):

NBCS, NBS, NCS, calf sera, calf serum

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.71

biological source

bovine (calf)

Quality Level

sterility

sterile-filtered

origin

New Zealand origin

technique(s)

cell culture | mammalian: suitable

impurities

≤35 EU/mL endotoxin

shipped in

dry ice

storage temp.

−20°C

General description

Serum is mostly required as a supplement to cell culture media. It contains macromolecules, carrier proteins, attachment and spreading factors, low molecular weight nutrients, and hormones and growth factors.
Newborn Bovine Serum (NBS) is collected from calves that are less than 10 days old. Although NBS is not as nutritionally rich as Fetal Bovine Serum (FBS), it can serve as a cost-effective substitute for FBS in many applications.

Biochem/physiol Actions

Newborn Calf Serum has been used in FACS (fluorescence-activated cell sorting) study for resuspension and dilution of cells.

Analysis Note

  • Endotoxin and hemoglobin tested
  • Tested for the presence of bacteria, virus, mycoplasma, and hemoglobin content
  • Triple filtered with 0.1 micron membrane under aseptic conditions

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Complete genome sequence and comparative genomic analysis of an emerging human pathogen, serotype V Streptococcus agalactiae.
Tettelin H et al.
Proceedings of the National Academy of Sciences of the USA, 99, 12391-12391 (2002)
Alternatives to the use of fetal bovine serum: serum-free cell culture.
Gstraunthaler G
ALTEX, 20, 275-275 (2003)
Ratnasekhar Ch et al.
Nature communications, 12(1), 377-377 (2021-01-17)
Circadian clocks coordinate mammalian behavior and physiology enabling organisms to anticipate 24-hour cycles. Transcription-translation feedback loops are thought to drive these clocks in most of mammalian cells. However, red blood cells (RBCs), which do not contain a nucleus, and cannot

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service