Skip to Content
Merck
All Photos(2)

Documents

D3877

Sigma-Aldrich

Diatomaceous earth

powder, suitable for most filtrations

Synonym(s):

Diatomaceous earth, calcined

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
11101527
PubChem Substance ID:
NACRES:
SB.52

form

powder

impurities

organic matter, free

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Diatomaceous earth can be used as an adsorbent for the adsorption of arsenic. It can also be used as a silica precursor to adsorb different groups of oil such as benzene, toluene, ethylbenzenes, p-xylenes, and o-xylenes.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Response surface optimization of oil removal using synthesized polypyrrole-silica polymer composite
Izevbekhai O, et al.
Molecules (Basel), 25, 4628-4628 (2020)
Miriam Eddyani et al.
Applied and environmental microbiology, 74(21), 6547-6553 (2008-09-09)
Buruli ulcer or Mycobacterium ulcerans disease occurs mainly in areas in proximity to standing or slowly running freshwater, habitats in which free-living amoebae occur. For this reason, a possible link between the habitat of M. ulcerans and free-living amoebae was
Competitive adsorption of As (III) and As (V) onto chitosan/diatomaceous earth adsorbent
Salih S, et al.
Journal of Environmental Chemical Engineering, 7, 103407-103407 (2019)
Bonhan Koo et al.
Scientific reports, 10(1), 14932-14932 (2020-09-12)
Cell-free nucleic acids (cfNAs) in liquid biopsy samples are emerging as important biomarkers for cancer diagnosis and monitoring, and for predicting treatment outcomes. Many cfNA isolation methods have been developed recently. However, most of these techniques are time-consuming, complex, require

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service