Skip to Content
Merck
All Photos(6)

Key Documents

799394

Sigma-Aldrich

Sodium bisulfite

anhydrous, free-flowing, Redi-Dri, ACS reagent

Synonym(s):

Sodium hydrogensulfite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
NaHSO3
CAS Number:
Molecular Weight:
104.06
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
anhydrous
form:
powder
Pricing and availability is not currently available.

grade

ACS reagent
anhydrous

Quality Level

product line

Redi-Dri

form

powder

quality

free-flowing

concentration

≥58.5% SO2

impurities

≤0.005% insolubles

pH

4.3 (10 g/L)

anion traces

chloride (Cl-): ≤0.02%

cation traces

Fe: ≤0.002%
heavy metals (as Pb): ≤0.001%

SMILES string

[Na+].OS([O-])=O

InChI

1S/Na.H2O3S/c;1-4(2)3/h;(H2,1,2,3)/q+1;/p-2

InChI key

DWAQJAXMDSEUJJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Sodium bisulfite is an inorganic sodium salt commonly used as a pH adjuster and reductant. It is used as a cheap catalyst for the formation of C-O and C-C bond.

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Supplementary Hazards

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hamid Mostafavi Abdolmaleky et al.
Methods in molecular biology (Clifton, N.J.), 448, 187-212 (2008-03-29)
Although there is evidence to link schizophrenia (SCZ) and bipolar disorder (BD) to genetic and environmental factors, specific individual or groups of genes/factors causative of the disease have been elusive to the research community. An understanding of the molecular aberrations
Synthesis of two new arsenolipids and their identification in fish.
Arroyo-Abad U, et al.
European Journal of Lipid Science and Technology, 118(3), 445-452 (2015)
Reaction of sodium bisulfite with uracil, cytosine, and their derivatives.
H Hayatsu et al.
Biochemistry, 9(14), 2858-2865 (1970-07-07)
Li-Hui Wang et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 19(5), 675-683 (2006-03-11)
The hedgehog (Hh) signaling pathway plays an important role in foregut development, and its activity is increased in various tumors, including those of the digestive tract. Our objective in the present study was to determine the pattern and extent of
Wei Zhou et al.
Journal of Korean medical science, 24(1), 97-103 (2009-03-10)
Epithelial membrane protein 3 (EMP3) is a trans-membrane signaling molecule with important roles in the regulation of apoptosis, differentiation and invasion of cancer cells, but the detailed is largely still unknown. We analyzed the mRNA levels and methylation statuses of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service