Skip to Content
Merck
All Photos(9)

Documents

746444

Sigma-Aldrich

Potassium bromide

greener alternative

anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KBr
CAS Number:
Molecular Weight:
119.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent
anhydrous

Quality Level

vapor pressure

<0.01 mmHg ( 20 °C)
1 mmHg ( 795 °C)

product line

Redi-Dri

Assay

≥99%

form

powder

quality

free-flowing

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

impurities

≤0.005% insolubles

pH

5.0-8.8 (25 °C, 5%)

mp

734 °C (lit.)

anion traces

bromate (BrO3-): ≤0.001%
chloride (Cl-): ≤0.2%
iodate (IO3-): ≤0.001%
iodide (I-): ≤0.001%
sulfate (SO42-): ≤0.005%

cation traces

Ba: ≤0.002%
Ca: ≤0.002%
Fe: ≤5 ppm
Mg: ≤0.001%
Na: ≤0.02%
heavy metals: ≤5 ppm (by ICP)

greener alternative category

SMILES string

[K+].[Br-]

InChI

1S/BrH.K/h1H;/q;+1/p-1

InChI key

IOLCXVTUBQKXJR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Application

May be used for removal of peripheral membrane proteins.

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hope E Baird-Heinz et al.
Journal of the American Veterinary Medical Association, 240(6), 705-715 (2012-03-03)
To critically evaluate and summarize available information on the safety of potassium bromide in dogs. Systematic review. 111 references reporting safety information relevant to potassium bromide published between 1938 and 2011. PubMed searches without date limitations were conducted with the
Hitesh Kulhari et al.
Nanomedicine : nanotechnology, biology, and medicine, 11(6), 1511-1520 (2015-05-03)
Docetaxel (DTX) is an anticancer drug that is used alone and in combination with other drugs to treat tumours. However, it suffers from the drawback of non-specific cytotoxicity. To improve the therapeutic potential of DTX, we report the synthesis of
Samy M El-Megharbel et al.
Chemico-biological interactions, 220, 169-180 (2014-07-08)
A new silver-carbocysteine (Ccy-Ag) complex [Ag2(Ccy)2(H2O)2] has been synthesized and characterized by using a combination of FTIR, Raman, molar conductivity, (1)H NMR, electronic spectra, thermal analyses, X-ray powder diffraction (XRD) and scanning electron microscopy (SEM). The infrared spectrum of Ccy-Ag
Dilip Devineni et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 88(2), 332-341 (2014-05-27)
Lyophilized formulations of keratinocyte growth factor-2 (KGF-2) were prepared with a range of disaccharide (sucrose or trehalose) and hydroxyethyl starch (HES) mass ratios. Protein degradation was assessed as a function of time of storage of the dried formulations at 40
Timothy D Heden et al.
Journal of applied physiology (Bethesda, Md. : 1985), 118(5), 624-634 (2014-12-30)
Abnormally elevated postprandial glucose and triacylglycerol (TAG) concentrations are risk factors for cardiovascular disease in type 2 diabetes. The most effective time to exercise to lower postprandial glucose and TAG concentrations is unknown. Thus the aim of this study was

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service