Skip to Content
Merck
All Photos(1)

Key Documents

Y0001649

Aripiprazole

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

7-{4-[4-(2,3-Dichlorophenyl)-1-piperazinyl]butoxy}-3,4-dihydro-2(1H)-quinolinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H27Cl2N3O2
CAS Number:
Molecular Weight:
448.39
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

aripiprazole

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Clc1c(cccc1N2CCN(CC2)CCCCOc3cc4c(cc3)CCC(=O)N4)Cl

InChI

1S/C23H27Cl2N3O2/c24-19-4-3-5-21(23(19)25)28-13-11-27(12-14-28)10-1-2-15-30-18-8-6-17-7-9-22(29)26-20(17)16-18/h3-6,8,16H,1-2,7,9-15H2,(H,26,29)

InChI key

CEUORZQYGODEFX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Aripiprazole EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel J Safer et al.
Journal of child and adolescent psychopharmacology, 23(4), 282-289 (2013-05-08)
This study aimed to: document the extent of the reduction of serum prolactin (PRL) levels induced by aripiprazole (ARI) treatment in children and adolescents, compare this effect by age group, and shed light on this phenomenon. PRL serum levels in
M Brunetti et al.
European review for medical and pharmacological sciences, 16(10), 1346-1354 (2012-10-30)
Aripiprazole is an atypical antipsychotic used for schizophrenia, manic and mixed episodes associated with bipolar I disorder and as adjunctive therapy for major depressive disorder. It functions as a partial agonist at dopamine D2 and 5-HT1A receptors, and as an
Farouq I Thabet et al.
Clinical neuropharmacology, 36(1), 29-30 (2013-01-22)
Aripiprazole is an atypical antipsychotic medication that is a partial dopamine D(2) and serotonin 5-hydroxytryptamine (1A) receptor agonist and 5-hydroxytryptamine (2A) receptor antagonist. It has a safer profile compared to other antipsychotic medications with regard to its effect on weight
Peter Paul De Deyn et al.
Expert opinion on pharmacotherapy, 14(4), 459-474 (2013-01-29)
Psychosis is a common and difficult to treat symptom in Alzheimer's disease (AD). It is a cause of diminished quality of life and caregiver distress. Atypical antipsychotics are frequently used for the treatment of dementia-related psychosis, despite FDA warnings because
Manit Srisurapanont et al.
Journal of psychiatric research, 62, 38-47 (2015-01-27)
Limited options are available for clozapine-resistant schizophrenia and intolerable side effects of clozapine. We conducted a systematic review of randomized-controlled trials (RCTs) to determine the efficacy and safety of aripiprazole augmentation of clozapine for schizophrenia. Electronic databases searched included PubMed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service