Skip to Content
Merck
All Photos(1)

Key Documents

PHR1212

Supelco

Anisole

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Methoxybenzene, Methyl phenyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H5
CAS Number:
Molecular Weight:
108.14
Beilstein:
506892
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1037011

vapor density

3.7 (vs air)

vapor pressure

10 mmHg ( 42.2 °C)

CofA

current certificate can be downloaded

autoignition temp.

887 °F

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.516 (lit.)

bp

154 °C (lit.)

mp

−37 °C (lit.)

density

0.995 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

COc1ccccc1

InChI

1S/C7H8O/c1-8-7-5-3-2-4-6-7/h2-6H,1H3

InChI key

RDOXTESZEPMUJZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Anisole is a volatile phenolic flavor compound that is reported to occur in cooked meat. It is obtained upon heating anisic acid with an excess of barium hydroxide.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA9025 in the slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

109.4 °F - closed cup

Flash Point(C)

43 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Laszlo Hollosi et al.
Journal of chromatography. A, 1218(1), 23-31 (2010-12-03)
European food legislation defines a set of 16 polycyclic aromatic hydrocarbons (PAHs) as of high concern for human health. The EU set contains structurally very similar PAHs with ring numbers between 4 and 6, and so raises some separation aspects
Xiaohui Jin et al.
Water research, 46(19), 6519-6530 (2012-10-20)
Second-order reaction rate constants of micropollutants with ozone (k(O3)) and hydroxyl radicals (k(OH)) are essential for evaluating their removal efficiencies from water during ozonation and advanced oxidation processes. Kinetic data are unavailable for many of the emerging micropollutants. Twenty-four micropollutants
Kohji Omata et al.
Journal of combinatorial chemistry, 12(4), 435-444 (2010-05-22)
To study the synergistic effect on the acidity and activity of Si-Al-Zr ternary oxide system, the mixture design of experiments was applied to prepare the ternary oxides by a sol-gel method, and rapid screening on dehydration of isopropanol and Friedel-Crafts
S Rivera et al.
Analytical and bioanalytical chemistry, 400(5), 1339-1346 (2011-03-08)
Various carotenoids were analyzed by ultra-high-pressure liquid chromatography with tandem mass spectrometry detection (UHPLC-MS/MS). Three different techniques to ionize the carotenoids were compared: electrospray ionization (ESI), atmospheric pressure chemical ionization (APCI) and atmospheric pressure photoionization (APPI). For all of the
Adam F Lee et al.
Dalton transactions (Cambridge, England : 2003), 39(43), 10473-10482 (2010-10-05)
Size-controlled, catalytically active PVP-stabilised Pd nanoparticles have been studied by operando liquid phase XAS during the Suzuki cross-coupling of iodonanisole and phenylboronic acid in MeOH-toluene using KOMe base. XAS reveals nanoparticles are stable to metal leaching throughout the reaction, with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service