Skip to Content
Merck
All Photos(1)

Key Documents

PHR1205

Supelco

Tributyl phosphate

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Tributyl phosphate, TBP, TBPA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3(CH2)3O)3PO
CAS Number:
Molecular Weight:
266.31
Beilstein:
1710584
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. Y0000279

vapor density

9.2 (vs air)

vapor pressure

27 mmHg ( 178 °C)
7.3 mmHg ( 150 °C)

API family

tributyl phosphate

CofA

current certificate can be downloaded

autoignition temp.

770 °F

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.424 (lit.)

bp

180-183 °C/22 mmHg (lit.)

mp

−79 °C (lit.)

density

0.979 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)

format

neat

storage temp.

2-30°C

SMILES string

CCCCOP(=O)(OCCCC)OCCCC

InChI

1S/C12H27O4P/c1-4-7-10-14-17(13,15-11-8-5-2)16-12-9-6-3/h4-12H2,1-3H3

InChI key

STCOOQWBFONSKY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tributyl phosphate is an industrial chemical, commonly used as a plasticizer and a solvent for the extraction of uranium and plutonium in nuclear industry.
Certified pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to in-house working standards.

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Ion Mobility: TWCCSN2 value of 164.9 Å2 [M+H]+ and 182.2 Å2 [M+Na]+

The collision cross section (CCS) measurement was provided by Waters Corporation, using the SYNAPT XS mass spectrometer.
For a description and overview of how ion mobility enables the measurement of the CCS of an ion visit ims.waters.com.
Further information on the SYNAPT XS mass spectrometer can be found on the IMS microsite and product webpage.

TWCCS measurements are expected to be within 2% of this reference value.

P/N PHR1205 is part of the Waters Extractables & Leachables UNIFI scientific library which can be downloaded from Waters Marketplace.

Footnote

To see an example of a Certificate of Analysis for this material enter P500205 in the slot below. This is an example certificate only and may not be the lot that you receive.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

293.0 °F - closed cup

Flash Point(C)

145 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Improved determination of tributyl phosphate degradation products (mono-and dibutyl phosphates) by ion chromatography
Dodi A and Verda G
Journal of Chromatography A, 920(1-2), 275-281 (2001)
T Burnouf et al.
Vox sanguinis, 104(1), 1-6 (2012-07-05)
A minipool solvent/detergent (S/D; 1% TnBP/1% Triton X-45; 31°C) process was developed for viral inactivation of plasma and cryoprecipitate used for transfusion. The goal of this study was to determine the rate and extent of inactivation of dengue virus (DENV)
C W Van Neste et al.
Analytical chemistry, 81(5), 1952-1956 (2009-02-04)
Despite its immediate applications, selective detection of trace quantities of surface adsorbed chemicals, such as explosives, without physically collecting the sample molecules is a challenging task. Standoff spectroscopic techniques offer an ideal method of detecting chemicals without using a sample
Domenico Santoro et al.
Environmental science & technology, 44(16), 6233-6241 (2010-08-14)
Advanced Oxidation Processes (AOPs) promoted by ultraviolet light are innovative and potentially cost-effective solutions for treating persistent pollutants recalcitrant to conventional water and wastewater treatment. While several studies have been performed during the past decade to improve the fundamental understanding
Muniasamy Neerathilingam et al.
Toxicology letters, 199(1), 10-16 (2010-08-07)
Tributyl phosphate (TBP) is a toxic organophosphorous compound widely used in many industrial applications, including significant usage in nuclear processing. The industrial application of this chemical is responsible for occupational exposure and environmental pollution. In this study, (1)H NMR-based metabonomics

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service