Skip to Content
Merck
All Photos(2)

Key Documents

NIST913B

Uric acid

NIST® SRM® 913b

Synonym(s):

2,6,8-Trihydroxypurine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4N4O3
CAS Number:
Molecular Weight:
168.11
Beilstein:
156158
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

crystalline (solid)

packaging

pkg of 10 g

manufacturer/tradename

NIST®

technique(s)

GC/MS: suitable
LC/MS: suitable
UV/Vis spectroscopy: suitable

mp

>300 °C (lit.)

application(s)

clinical testing

format

neat

SMILES string

O=C1NC(=O)C2=C(N1)NC(=O)N2

InChI

1S/C5H4N4O3/c10-3-1-2(7-4(11)6-1)8-5(12)9-3/h(H4,6,7,8,9,10,11,12)

InChI key

LEHOTFFKMJEONL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This uric acid standard reference material (SRM) is a chemical with known purity. A unit of SRM 913b consists of one bottle containing 10 g of crystalline uric acid.

NIST913B_Cert

NIST913B_SDS

Application

This uric acid is intended primarily for use in the calibration and standardization of procedures for uric acid determinations employed in clinical analysis. It is also used for routine critical evaluation of the daily working standards used in these procedures.

Other Notes

  • Refer to the NIST certificate for details on expiry, storage, and usage.
  • Notes for biomaterials, disposal, and transport information are available in SDS.
Certified for the analytes listed below.
Uric Acid (C5H4N4O3)

See certificate for values and more details at nist.gov/SRM.

Preparation Note

  • The bottle must be stored at temperatures between 20 °C and 25 °C and protected from excessive light and moisture.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Core Concepts in Parenchymal Kidney Disease (2013)
Towards the physiological function of uric acid.
Becker F B.
Free Radical Biology & Medicine, 14(6), 615-631 (1993)
Primer on Kidney Diseases, Volume 953 (2005)
Measurement of serum uric acid by isotope dilution liquid chromatography tandem mass spectrometry: Modification of a candidate reference
Jun Y, et al.
International Journal of Mass Spectrometry, 482, 116911-116911 (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service