Skip to Content
Merck
All Photos(1)

Key Documents

L0400000

Levodopa

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

3,4-Dihydroxy-L-phenylalanine, 3-(3,4-Dihydroxyphenyl)-L-alanine, L-3-Hydroxytyrosine, L-DOPA, Levodopa

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2C6H3CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
197.19
Beilstein:
2215169
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

levodopa

manufacturer/tradename

EDQM

mp

276-278 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

N[C@@H](Cc1ccc(O)c(O)c1)C(O)=O

InChI

1S/C9H11NO4/c10-6(9(13)14)3-5-1-2-7(11)8(12)4-5/h1-2,4,6,11-12H,3,10H2,(H,13,14)/t6-/m0/s1

InChI key

WTDRDQBEARUVNC-LURJTMIESA-N

Gene Information

human ... DRD3(1814)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Levodopa EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pol Nadal-Jimenez et al.
Journal of bacteriology, 196(14), 2681-2690 (2014-05-13)
The iron binding siderophore pyoverdine constitutes a major adaptive factor contributing to both virulence and survival in fluorescent pseudomonads. For decades, pyoverdine production has allowed the identification and classification of fluorescent and nonfluorescent pseudomonads. Here, we demonstrate that PvdP, a
Nicolas Morin et al.
Experimental neurology, 256, 105-116 (2013-01-31)
The treatment of motor symptoms of Parkinson disease (PD) with the dopamine (DA) precursor, l-3,4-dihydroxyphenylalanine (l-DOPA) introduced 50years ago still remains a very effective medication. However, involuntary movements termed l-DOPA-induced dyskinesias (LID) appear in the vast majority of PD patients
Elahe Molaakbari et al.
The Analyst, 139(17), 4356-4364 (2014-07-12)
A novel carbon paste electrode modified with ZnO nanorods and 5-(4'-amino-3'-hydroxy-biphenyl-4-yl)-acrylic acid (3,4'-AAZCPE) was fabricated. The electrochemical study of the modified electrode, as well as its efficiency for the electrocatalytic oxidation of levodopa, is described. The electrode was employed to
M Purschke et al.
The British journal of dermatology, 172(4), 1021-1028 (2014-10-14)
Epidermal suction blister grafts are an effective treatment for chronic wounds or vitiligo, but this treatment is time consuming and limited to small areas. To compare two novel strategies to create fractional epidermal grafts. Epidermal blisters were raised from fresh
Vincent J J Odekerken et al.
Neurology, 84(13), 1355-1361 (2015-03-01)
To assess the neuropsychological outcome 12 months after bilateral deep brain stimulation (DBS) of the globus pallidus pars interna (GPi) or subthalamic nucleus (STN) for advanced Parkinson disease. We randomly assigned patients to receive either GPi DBS or STN DBS.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service