Skip to Content
Merck
All Photos(1)

Key Documents

K2000000

Ketoprofen

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-(3-Benzoylphenyl)propionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14O3
CAS Number:
Molecular Weight:
254.28
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

ketoprofen

manufacturer/tradename

EDQM

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CC(C(O)=O)c1cccc(c1)C(=O)c2ccccc2

InChI

1S/C16H14O3/c1-11(16(18)19)13-8-5-9-14(10-13)15(17)12-6-3-2-4-7-12/h2-11H,1H3,(H,18,19)

InChI key

DKYWVDODHFEZIM-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ketoprofen EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Non-steroidal anti-inflammatory compound that is selective for COX-1.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Ikram Ullah Khan et al.
International journal of pharmaceutics, 441(1-2), 809-817 (2012-12-26)
Microchannels based microfluidic systems are able to obtain monodispersed microparticles but are limited by cost, time and channel clogging. We succeeded in on the fly encapsulation of high ketoprofen contents in acrylate-based copolymer microbeads by environment friendly UV induced free
Yan-Ping Duan et al.
The Science of the total environment, 447, 267-273 (2013-02-09)
The occurrence and multi-phase distribution of five pharmaceutical compounds were investigated in an urban wastewater treatment plant (WWTP) receiving river by analysis of pharmaceuticals in sediment, particulate matter, conventional dissolved phase (>0.7 μm), colloidal phase (5 kDa to 0.7 μm)
Zeting Yuan et al.
International journal of pharmaceutics, 446(1-2), 191-198 (2013-02-21)
Chitosan (CS) grafted with β-cyclodextrin (CD-g-CS) nanoparticles as a new carrier for poorly water-soluble drugs has been developed. The CD-g-CS polymer is readily synthesized from chitosan and mono-6-deoxy-6-(p-toluenesulfonyl)-β-cyclodextrin. Three different degrees of substitution (DS) of β-cyclodextrin (β-CD) on CD-g-CS (9.6
Ariane Rennó Brogliato et al.
Shock (Augusta, Ga.), 38(6), 620-629 (2012-11-13)
The mechanism of immunosuppression induced by severe sepsis is not fully understood. The production of prostaglandin E2 (PGE2) during sepsis is well known, but its role in long-term consequences of sepsis has not been explored. The current study evaluates the
F Jamali et al.
Clinical pharmacokinetics, 19(3), 197-217 (1990-09-01)
Ketoprofen, a potent nonsteroidal anti-inflammatory drug (NSAID) of the 2-arylpropionic acid class, has been used clinically for over 15 years in Europe, and has recently been introduced in the United States. Although it possesses a chiral centre, with only the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service