Skip to Content
Merck
All Photos(4)

Key Documents

D3252

Sigma-Aldrich

o-Dianisidine dihydrochloride

≥95%

Synonym(s):

3,3′-Dimethoxybenzidine dihydrochloride, Fast Blue B

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H16N2O2 · 2HCl
CAS Number:
Molecular Weight:
317.21
Beilstein:
3917996
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47
Pricing and availability is not currently available.

Quality Level

Assay

≥95%

form

powder

mp

268 °C

solubility

H2O: soluble 25 mg/mL

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

Cl.Cl.COc1cc(ccc1N)-c2ccc(N)c(OC)c2

InChI

1S/C14H16N2O2.2ClH/c1-17-13-7-9(3-5-11(13)15)10-4-6-12(16)14(8-10)18-2;;/h3-8H,15-16H2,1-2H3;2*1H

InChI key

UXTIAFYTYOEQHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Specificity

o-Dianisidine (3,3′-dimethoxybenzidine) is a peroxidase substrate suitable for use in ELISA procedures. This substrate produces a soluble end product that is yellow-orange in color and can be read spectrophotometrically at 405 nm. The reaction may be stopped with 5 M HCl.

Application

o-Dianisidine (3,3′-dimethoxybenzidine) has been used as a component of GO (glucose oxidase) reagent, which is used for measuring the glucose content.[1] It has also been used as a substrate to measure myeloperoxidase activity.[2]

Warning

Probably carcinogenic.

Physical form

crystalline

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Eye Dam. 1 - Skin Corr. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luminol-enhanced chemiluminescence induced in peripheral blood-derived human phagocytes: obligatory requirement of myeloperoxidase exocytosis by monocytes.
Albrecht D and Jungi TW
Journal of Leukocyte Biology, 54, 300-300 (1993)
Interspecies interactions determine the impact of the gut microbiota on nutrient allocation in Drosophila melanogaster.
Newell PD and Douglas AE
Applied and Environmental Microbiology, 80, 788-788 (2014)
Peter M Meggyesy et al.
Molecules (Basel, Switzerland), 25(21) (2020-10-31)
The therapeutic utility of the copper ionophore disulfiram was investigated in a diet-induced obesity mouse model (C57BL/6J background), both through administration in feed (0.05 to 1% (w/w)) and via oral gavage (150 mg/kg) for up to eight weeks. Mice were
Zhiquan Chen et al.
International journal of molecular sciences, 21(15) (2020-07-28)
Intestinal ischemia reperfusion injury (IRI) is an inherent, unavoidable event of intestinal transplantation, contributing to allograft failure and rejection. The inflammatory state elicited by intestinal IRI is characterized by heightened leukocyte recruitment to the gut, which is amplified by a
Rémy Ricoux et al.
Bioconjugate chemistry, 19(4), 899-910 (2008-03-08)
To develop artificial hemoproteins that could lead to new selective oxidation biocatalysts, a strategy based on the insertion of various iron-porphyrin cofactors into Xylanase A (Xln10A) was chosen. This protein has a globally positive charge and a wide enough active

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service