Skip to Content
Merck
All Photos(1)

Key Documents

D0160000

Deferoxamine mesylate salt

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

DFOM, Deferoxamine methanesulfonate salt, Desferrioxamine mesylate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H48N6O8 · CH4O3S
CAS Number:
Molecular Weight:
656.79
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

deferoxamine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CS(O)(=O)=O.CC(=O)N(O)CCCCCNC(=O)CCC(=O)N(O)CCCCCNC(=O)CCC(=O)N(O)CCCCCN

InChI

1S/C25H48N6O8.CH4O3S/c1-21(32)29(37)18-9-3-6-16-27-22(33)12-14-25(36)31(39)20-10-4-7-17-28-23(34)11-13-24(35)30(38)19-8-2-5-15-26;1-5(2,3)4/h37-39H,2-20,26H2,1H3,(H,27,33)(H,28,34);1H3,(H,2,3,4)

InChI key

IDDIJAWJANBQLJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Deferoxamine mesylate salt EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

An iron chelator used often in the studies of cell proliferation and apoptosis. Also reported to induce p53.
An iron chelator used often in the studies of cell proliferation and apoptosis. Has been shown to have anti-proliferative effects on vascular smooth muscle cells in vitro and in vivo and to arrest cells in the G1 phase. Also reported to induce p53. Induces apoptosis in HL-60 cells by chelating iron. After 48 hrs treatment with 1μM deferoxamine, DNA fragmentation was apparent. Cells treated with 0.1 μM deferoxamine for as little as 24 hours were committed to apoptosis; by 48 hrs nuclear collapse was observed. In some studies it has been shown to have antioxidant properties and to protect cells against H2O2-induced damage.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xueping Guo et al.
PloS one, 9(4), e94156-e94156 (2014-04-17)
Hyaluronidases are a family of enzymes that degrade hyaluronic acid (hyaluronan, HA) and widely used in many fields. A hyaluronidase producing bacteria strain was screened from the air. 16S ribosomal DNA (16S rDNA) analysis indicated that the strain belonged to
Cindy A Ryan et al.
Toxicological sciences : an official journal of the Society of Toxicology, 178(1), 88-103 (2020-10-24)
A peptide reactivity assay with an activation component was developed for use in screening chemicals for skin sensitization potential. A horseradish peroxidase-hydrogen peroxide (HRP/P) oxidation system was incorporated into the assay for characterizing reactivity of hapten and pre-/prohapten sensitizers. The
Yoshiaki Tsuji
PloS one, 15(7), e0235563-e0235563 (2020-07-10)
Western blotting has been widely used for investigation of protein expression, posttranslational modifications, and interactions. Because western blotting usually involves heat-denaturation of samples prior to gel loading, clarification of detailed procedures for sample preparation have been omitted or neglected in
Meredith Corley et al.
Molecular cell, 80(5), 903-914 (2020-11-27)
Discovering the interaction mechanism and location of RNA-binding proteins (RBPs) on RNA is critical for understanding gene expression regulation. Here, we apply selective 2'-hydroxyl acylation analyzed by primer extension (SHAPE) on in vivo transcripts compared to protein-absent transcripts in four human
Joao Fonseca et al.
Oncology letters, 17(1), 697-705 (2019-01-19)
Resveratrol (RES) is a polyphenol produced by certain plant species that has been well studied due to its ability to slow the growth of cancer cells. In numerous cell types and tissues, RES has been demonstrated to promote mitochondrial biogenesis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service