Skip to Content
Merck
All Photos(1)

Key Documents

C1915

Supelco

Chloropyramine hydrochloride

analytical standard, for drug analysis

Synonym(s):

N-p-Chlorobenzyl-N′,N′-dimethyl-N-(2-pyridyl)ethylenediamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H20ClN3 · HCl
CAS Number:
Molecular Weight:
326.26
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

SMILES string

Cl.CN(C)CCN(Cc1ccc(Cl)cc1)c2ccccn2

InChI

1S/C16H20ClN3.ClH/c1-19(2)11-12-20(16-5-3-4-10-18-16)13-14-6-8-15(17)9-7-14;/h3-10H,11-13H2,1-2H3;1H

InChI key

VEYWWAGBHABATA-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Budihna et al.
Pflugers Archiv : European journal of physiology, 431(6 Suppl 2), R217-R218 (1996-01-01)
In isolated rat hearts effects of chloropyramine (CP), histamine H1 antagonist, and famotidine (FA), H2 antagonist, upon two different myocardial injuries, ischaemia-reperfusion and hypoxia-reoxygenation were studied. In both types of injury the effects of drugs were seen mainly during reperfusion
[Application of aqueous Eudragit dispersions to the preparation of controlled release oral antihistamine dosage forms].
A Dévay et al.
Acta pharmaceutica Hungarica, 58(4), 166-172 (1988-07-01)
[Toxico-allergic form of erythema multiforme exudativum caused by suprastin].
S M Fedorov et al.
Vestnik dermatologii i venerologii, (9)(9), 50-51 (1986-01-01)
D B Beleslin et al.
Neuropharmacology, 26(8), 1157-1165 (1987-08-01)
The emetic action of noradrenaline was investigated in unanesthetized cats, after it was injected into the cerebral ventricles through chronically implanted cannulae. Intracerebroventricular injection of noradrenaline produced dose-dependent and shortlasting emesis, which was abolished after ablation of the area postrema.
K I Bender et al.
Farmakologiia i toksikologiia, 52(3), 66-71 (1989-05-01)
In the experiments on male albino rats absorption of dimedrol, pipolphen and suprastin from the small intestine was studied in vitro by the method of "turned sacks". It was shown that immunization of the animals with ovalbumin in combination with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service