Skip to Content
Merck
All Photos(1)

Key Documents

91379

Supelco

Boron trichloride solution

12% in methanol, for GC derivatization, LiChropur

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
BCl3
CAS Number:
Molecular Weight:
117.17
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.22

grade

for GC derivatization

Quality Level

form

liquid

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Acylations

concentration

11.0-13.0% (alkalimetric)
12% in methanol

technique(s)

gas chromatography (GC): suitable

density

0.875-0.895 g/mL at 20 °C

storage temp.

−20°C

SMILES string

ClB(Cl)Cl

InChI

1S/BCl3/c2-1(3)4

InChI key

FAQYAMRNWDIXMY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Boron trichloride solution is an esterification reagent. It is a powerful acidic catalyst for the esterification of fatty acids for GC analysis.

Application

Learn more in the Product Information
Borontrichloride solution is used as a derivatization reagent for reliable andaccurate identification and quantification of fatty acids mixtures by GC-FID.
For preparing methyl esters of fatty acids and for transesterification of triglycerides.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Eye Dam. 1 - Flam. Liq. 2 - Repr. 1B - Skin Corr. 1B - STOT SE 1

Target Organs

Eyes,Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gas chromatography determination of fatty acids in the human erythrocyte membranes-A review
Bystricka Z, et al.
Prostaglandins, Leukotrienes, and Essential Fatty Acids, 115, 35-40 (2016)
Ummara Butt et al.
Drug delivery and translational research, 6(6), 722-734 (2016-10-22)
Fatty acids (FAs) are used by many organisms as defence mechanism against virulent bacteria. The high safety profile and broad spectrum of activity make them potential alternatives to currently used topical antibiotics for the treatment of eye infections in neonates.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service