Skip to Content
Merck
All Photos(1)

Documents

74820

Supelco

Octane

analytical standard

Synonym(s):

n-Octane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6CH3
CAS Number:
Molecular Weight:
114.23
Beilstein:
1696875
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

3.9 (vs air)

vapor pressure

11 mmHg ( 20 °C)

Assay

≥99.7% (GC)

autoignition temp.

428 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

6.5 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.397
n20/D 1.398 (lit.)

bp

125-127 °C (lit.)

mp

−57 °C (lit.)

density

0.703 g/mL at 25 °C (lit.)

application(s)

petroleum

format

neat

SMILES string

CCCCCCCC

InChI

1S/C8H18/c1-3-5-7-8-6-4-2/h3-8H2,1-2H3

InChI key

TVMXDCGIABBOFY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 3

1 of 3

David L Cheung
Langmuir : the ACS journal of surfaces and colloids, 28(23), 8730-8736 (2012-05-18)
Hydrophobins are small, amphiphilic proteins expressed by strains of filamentous fungi. They fulfill a number of biological functions, often related to adsorption at hydrophobic interfaces, and have been investigated for a number of applications in materials science and biotechnology. In
Mariela Ivanova et al.
Electrophoresis, 24(6), 992-998 (2003-03-27)
Microemulsion electrokinetic chromatography was applied for the separation of levetiracetam from other antiepileptic drugs (primidone, phenobarbital, phenytoin, lamotrigine and carbamazepine) that are potentially coadministered in therapy of patients. The influence of the composition of the microemulsion system (with sodium dodecyl
Oluwafemi A Olaofe et al.
Microbial cell factories, 12, 8-8 (2013-01-29)
Biocatalyst improvement through molecular and recombinant means should be complemented with efficient process design to facilitate process feasibility and improve process economics. This study focused on understanding the bioprocess limitations to identify factors that impact the expression of the terminal
Jian Xie et al.
Organic letters, 15(4), 868-870 (2013-02-09)
The use of a powerful intramolecular [4 + 2]/[3 + 2] cycloaddition cascade of an 1,3,4-oxadiazole in the divergent total synthesis of kopsinine (1), featuring an additional unique SmI(2)-promoted transannular cyclization reaction for formation of the bicyclo[2.2.2]octane central to its
K E Kramer et al.
Journal of chromatography. B, Biomedical sciences and applications, 760(1), 27-36 (2001-08-28)
An efficient and inexpensive screening technique for the simultaneous clean-up, extraction, and derivatization of 11-nor-delta9-tetrahydrocannabinol-9-carboxylic acid (THC-COOH) from urine has been developed. Using the principles of solvent microextraction in the form of a 20-microl volume of solvent placed inside a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service