Skip to Content
Merck
All Photos(1)

Key Documents

61044

Millipore

Casein Peptone

Enzymatic digest of casein from bovine milk, suitable for biotechnology and microbiology

Synonym(s):

Peptone from casein, Peptone from casein Plus, tryptic digest, Tryptone Plus

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
41106212
NACRES:
NA.85

product name

Tryptone Plus, for biotechnological purposes

biological source

bovine milk

Quality Level

grade

for biotechnological purposes

form

powder

composition

amino-nitrogen, ≥4%
total nitrogen (N), ≥12%

ign. residue

≤22%

loss

≤8.0% loss on drying

pH

6.2-7.2 (25 °C)

solubility

H2O: 2%, clear, faintly yellow to brownish-yellow

application(s)

food and beverages
microbiology

Looking for similar products? Visit Product Comparison Guide

General description

Source of organic nitrogen and growth factors recommended in media for analytical microbiology and industrial fermentation. This casein peptone is highly soluble.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of medium components on bacteriocin production by Lactobacillus pentosus ST151BR, a strain isolated from beer produced by the fermentation of maize, barley and soy flour.
Todorov, S. D., and L. M. T. Dicks.
World Journal of Microbiology & Biotechnology, 20 (6), 643-650 (2004)
Studies on transformation of Escherichia coli with plasmids.
Hanahan, Douglas.
Journal of Molecular Biology, 166 (4), 557-580 (1983)
Thermomicrobium, a new genus of extremely thermophilic bacteria
JACKSON, THOMAS J., ROBERT F. RAMALEY, and WARREN G. MEINSCHEIN
International Journal of Systematic and Evolutionary Microbiology, 23 (1), 28-36 (1973)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service