Skip to Content
Merck
All Photos(1)

Documents

45945

Supelco

Isopropylamine

anhydrous, analytical standard

Synonym(s):

2-Aminopropane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHNH2
CAS Number:
Molecular Weight:
59.11
Beilstein:
605259
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

vapor density

2.04 (vs air)

vapor pressure

9.2 psi ( 20 °C)

autoignition temp.

755 °F

quality

anhydrous

shelf life

limited shelf life, expiry date on the label

expl. lim.

10.4 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.374 (lit.)

bp

33-34 °C (lit.)

density

0.688 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CC(C)N

InChI

1S/C3H9N/c1-3(2)4/h3H,4H2,1-2H3

InChI key

JJWLVOIRVHMVIS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 1 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

<-13.0 °F - closed cup

Flash Point(C)

<= -25 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hsin-Ling Lee et al.
Clinical toxicology (Philadelphia, Pa.), 47(7), 651-658 (2009-08-12)
Most of the glyphosate-surfactant herbicides (GlySH) are formulated commercial products containing isopropylamine (IPA) salt of glyphosate (IPAG), variable amount of a surfactant, and water. Although glyphosate is only slightly toxic to rats, ingestion of GlySH may lead to severe effects
Jacek Lipok et al.
Ecotoxicology and environmental safety, 73(7), 1681-1688 (2010-09-04)
The toxicity of commercial formulation of Roundup® 360 SL, widely used, nonselective herbicide and its main constituents, glyphosate (PMG), equimolar (1:1) isopropylamine salt of glyphosate (GIPA) and isopropylamine (IPA) was examined towards eight aquatic microphotoautotrophs; seven cyanobacterial strains representing either
Katrijn De Klerck et al.
Journal of chromatography. A, 1234, 72-79 (2011-12-07)
In chiral supercritical fluid chromatography (SFC), mobile-phase additives are often used to improve enantioseparations and peak shapes. An acidic or basic additive is chosen, depending on the nature of the compound. This work highlights the simultaneous use of the acidic
Gabriel Vallejos et al.
Bioorganic & medicinal chemistry, 13(14), 4450-4457 (2005-05-24)
The in vitro monoamine oxidase inhibitory (MAOI) activities of 11 heteroarylisopropylamines vis-à-vis MAO-A and MAO-B were described and interpreted in terms of possible interactions with the enzyme active site. Molecular dynamics simulations allowed a comparison between the most active MAO-A
Bambang Veriansyah et al.
Journal of hazardous materials, 124(1-3), 119-124 (2005-06-09)
Supercritical water oxidation can effectively destroy a large variety of high-risk wastes resulting from munitions demilitarization and complex industrial chemical. An important design consideration in the development of supercritical water oxidation is the information on the oxidation rate. In this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service