Skip to Content
Merck
All Photos(1)

Key Documents

38292

Supelco

Diisopropylamine

analytical standard

Synonym(s):

DIPA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHNHCH(CH3)2
CAS Number:
Molecular Weight:
101.19
Beilstein:
605284
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

3.5 (vs air)

vapor pressure

50 mmHg ( 20 °C)

Assay

≥99.5% (GC)

autoignition temp.

599 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

8.5 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.392 (lit.)
n20/D 1.392

bp

84 °C (lit.)

mp

−61 °C (lit.)

solubility

H2O: soluble 110 g/L at 25 °C (completely)

density

0.722 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

CC(C)NC(C)C

InChI

1S/C6H15N/c1-5(2)7-6(3)4/h5-7H,1-4H3

InChI key

UAOMVDZJSHZZME-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diisopropylamine, an aliphatic secondary amine, is a well-known precursor of lithium diisopropylamide (LDA), which is a hindered, strong, non-nucleophilic base utilized in various organic reactions.

Application

Diisopropylamine may be used as an analytical standard for the determination of the analyte in air samples by high performance liquid chromatography (HPLC) technique.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

7.8 °F - closed cup

Flash Point(C)

-13.45 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of aliphatic primary and secondary amines and polyamines in air by high-performance liquid chromatography
Simon P and Lemacon C
Analytical Chemistry, 59(3), 480-484 (1987)
Reductions of. alpha.-substituted ketones by lithium diisopropylamide
Kowalski C, et al.
The Journal of Organic Chemistry, 43(13), 2601-2608 (1978)
Koichi Mikami et al.
Chemical record (New York, N.Y.), 6(1), 1-11 (2006-02-14)
The difficulty as well as the significance of the direct generation of metal enolates of alpha-CF(3) ketones cannot be easily understood by chemists unfamiliar with F. In sharp contrast to the sunny side of non-F, hydrocarbon chemistry, F chemistry has
Decoupling deprotonation from metalation: Thia-Fries rearrangement.
Alan M Dyke et al.
Angewandte Chemie (International ed. in English), 47(27), 5067-5070 (2008-05-28)
Songping Liao et al.
Journal of the American Chemical Society, 125(49), 15114-15127 (2003-12-05)
Lithium diisopropylamide-mediated lithiations of N-alkyl ketimines derived from cyclohexanones reveal that simple substitutions on the N-alkyl side chain and the 2-position of the cyclohexyl moiety afford a 60,000-fold range of rates. Detailed rate studies implicate monosolvated monomers at the rate-limiting

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service