Skip to Content
Merck
All Photos(2)

Key Documents

38176

Supelco

Thiamethoxam-d3

PESTANAL®, analytical standard

Synonym(s):

3-(2-Chloro-5-thiazolylmethyl)tetrahydro-5-methyl-d3-N-nitro-4H-1,3,5-oxadiazin-4-imine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8D3H7ClN5O3S
CAS Number:
Molecular Weight:
294.73
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

mass shift

M+3

SMILES string

[2H]C([2H])([2H])N1COCN(Cc2cnc(Cl)s2)\C1=N\[N+]([O-])=O

InChI

1S/C8H10ClN5O3S/c1-12-4-17-5-13(8(12)11-14(15)16)3-6-2-10-7(9)18-6/h2H,3-5H2,1H3/b11-8+/i1D3

InChI key

NWWZPOKUUAIXIW-VZOBLPKOSA-N

General description

Find more information here: Neonicotinoids

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Thiamethoxam-d3 has played a role of an internal standard, in the determination of several neonicotinoid insecticides present in bee pollen, using ultra-high performance liquid chromatography (UHPLC) coupled with mass spectrometry(MS).

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Sol. 1 - Repr. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hua Zhang et al.
Environmental research, 199, 111175-111175 (2021-05-09)
Neonicotinoid insecticides (NEOs) are widely used for pest control worldwide. The profile of NEOs in paired urine and indoor dust has not yet been reported in China. In this study, 40 paired samples (i.e., 160 urine and 40 indoor dust)
Daniel B Pickford et al.
Environmental toxicology and chemistry, 37(4), 1040-1050 (2017-11-07)
Thiamethoxam is a widely used neonicotinoid insecticide that has been detected in surface water monitoring programs in North America and Europe. This has led to questions about its toxicity to nontarget insects, specifically those with an aquatic life stage. To
C Lobson et al.
The Science of the total environment, 637-638, 1150-1157 (2018-05-29)
Thiamethoxam is a neonicotinoid insecticide that can reach wetlands in agro-ecosystems through runoff. The fate and effects of thiamethoxam on non-target organisms in shallow wetland ecosystems have not been well characterized. To this end, a mesocosm study was conducted with
Lucas M Morrison et al.
Journal of AOAC International, 101(6), 1940-1947 (2018-05-23)
Background: Neonicotinoids are among the most widely used insecticides. Recently, there has been concern associated with unintended adverse effects on honeybees and aquatic invertebrates at low parts-per-trillion levels. Objective: There is a need for LC-MS/MS methods that are capable of
Junjie Zhang et al.
Talanta, 170, 392-398 (2017-05-16)
Extensive use of neonicotinoid insecticides has raised great concerns about their ecological risk. A reliable method to measure trace neonicotinoids in complicated aquatic environment is a premise for assessing their aquatic risk. To effectively remove matrix interfering substances from field

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service