Skip to Content
Merck
All Photos(1)

Documents

35562

Supelco

Dalapon

PESTANAL®, analytical standard

Synonym(s):

2,2-Dichloropropionic acid, Dalapon

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CCl2CO2H
CAS Number:
Molecular Weight:
142.97
Beilstein:
1750149
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.454 (lit.)

bp

90-92 °C/14 mmHg (lit.)

density

1.389 g/mL at 25 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

CC(Cl)(Cl)C(O)=O

InChI

1S/C3H4Cl2O2/c1-3(4,5)2(6)7/h1H3,(H,6,7)

InChI key

NDUPDOJHUQKPAG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dalapon is a small, water soluble, organochlorine selective herbicide and plant growth regulator used to control perennial grasses. It is mainly used on food crops, including sugarcane and sugar beets.

Application

Dalapon may be used as an analytical reference standard for the determination of the analyte in water and soil samples by liquid chromatography with tandem mass spectrometric detection based on dispersive solid phase extraction and liquid-solid disk extraction and capillary gas chromatography, respectively
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of acid herbicides in aqueous samples by liquid?solid disk extraction and capillary gas chromatography.
Hodgeson J, et al.
Journal of Chromatography A, 659(2), 395-401 (1994)
Pesticide Profiles: Toxicity, Environmental Impact, and Fate (1997)
Rapid determination of acid herbicides in soil by liquid chromatography with tandem mass spectrometric detection based on dispersive solid phase extraction.
Kaczynski P, et al.
Talanta, 152, 127-136 (2016)
Darryl W Hawker et al.
Journal of environmental monitoring : JEM, 13(2), 252-256 (2010-12-18)
Salts of 2,2-dichloropropionic acid, such as dalapon, are well known as herbicides and are regulated as such in potable water in Australia and elsewhere. It is also an identified disinfection by-product (DBP), but little is known about the compound's formation
Robert J Wells et al.
Journal of agricultural and food chemistry, 50(12), 3366-3374 (2002-05-30)
Comparison of quantitative NMR spectroscopy (QNMR) with chromatographic methods such as gas chromatography (GC) or high-pressure liquid chromatography (HPLC) for the determination of the purity of and impurities in technical grade agrochemicals, 2,4-dichlorophenoxyacetic acid (2,4-D), 1, and Dalapon sodium (sodium

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service