Skip to Content
Merck
All Photos(2)

Key Documents

35361

Supelco

Diclofenac-(acetophenyl ring-13C6) sodium salt hemi(nonahydrate)

VETRANAL®, analytical standard

Synonym(s):

[2-(2,6-Dichlorophenylamino)phenyl13C6]acetic acid sodium salt hemi(nonahydrate)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13C6C8H10Cl2NNaO2 · 4.5H2O
CAS Number:
Molecular Weight:
405.16
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

mass shift

M+12

SMILES string

O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]C(=O)C[13c]1[13cH][13cH][13cH][13cH][13c]1Nc2c(Cl)cccc2Cl.[O-]C(=O)C[13c]3[13cH][13cH][13cH][13cH][13c]3Nc4c(Cl)cccc4Cl

InChI

1S/2C14H11Cl2NO2.2Na.9H2O/c2*15-10-5-3-6-11(16)14(10)17-12-7-2-1-4-9(12)8-13(18)19;;;;;;;;;;;/h2*1-7,17H,8H2,(H,18,19);;;9*1H2/q;;2*+1;;;;;;;;;/p-2/i2*1+1,2+1,4+1,7+1,9+1,12+1;;;;;;;;;;;

InChI key

PRLOWEIBESKIQR-IJYJQIEZSA-L

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Magda Caban et al.
Analytical methods : advancing methods and applications, 12(30), 3854-3864 (2020-07-24)
The study aimed to show the limitations and advantages of the use of stable isotope labeled internal standards (SILISs) for the quantification of small polar compounds (ibuprofen, diclofenac, metoprolol, bisphenol A, 17β-estradiol) in water samples by GC/MS with selected ion

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service