Skip to Content
Merck
All Photos(1)

Documents

32532

Supelco

Isopyrazam

PESTANAL®, analytical standard, mixture of isomers

Synonym(s):

3-(Difluoromethyl)-1-methyl-N-[1,2,3,4-tetrahydro-9-(1-methylethyl)-1,4-methanonaphthalen-5-yl]-1H-pyrazole-4-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H23F2N3O
CAS Number:
Molecular Weight:
359.41
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC(C)C1C2CCC1c3c(NC(=O)c4cn(C)nc4C(F)F)cccc23

InChI

1S/C20H23F2N3O/c1-10(2)16-12-7-8-13(16)17-11(12)5-4-6-15(17)23-20(26)14-9-25(3)24-18(14)19(21)22/h4-6,9-10,12-13,16,19H,7-8H2,1-3H3,(H,23,26)

InChI key

XTDZGXBTXBEZDN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Isopyrazams are categorized under the class of succinate-dehydrogenase-inhibitor (SDHI) fungicides that are applied on a variety of fruits and vegetables.

Application

Isopyrazam may be used as an analytical reference standard for the determination of the analyte in:
  • Honeybees by dispersive solid-phase extraction (dSPE), followed by liquid and gas chromatography combined with tandem mass spectrometry (LC-MS/MS and GC-MS/MS) operating in electron ionization (EI) and multiple reaction monitoring (MRM) modes of detection.
  • Bee pollen samples by GC-MS/MS in single reaction monitoring (SRM) mode of detection.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Repr. 1B - Skin Sens. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A sensitive and efficient method for routine pesticide multiresidue analysis in bee pollen samples using gas and liquid chromatography coupled to tandem mass spectrometry.
Vazquez PP, et al.
Journal of Chromatography A, 1426(14), 161-173 (2015)
Multi-residue method for the determination of pesticides and pesticide metabolites in honeybees by liquid and gas chromatography coupled with tandem mass spectrometry?Honeybee poisoning incidents.
Kiljanek T, et al.
Journal of Chromatography A, 1435(14), 100-114 (2016)
Khuram Shahzad Ahmad et al.
Journal of basic microbiology, 60(6), 484-493 (2020-04-22)
The current research is a pioneer in the evaluation of isopyrazam biodegradation, which has been performed utilizing soil-isolated microbes. Biodisintegrative assays of pure fungal strains, namely Aspergillus flavus (AF), Penicillium chrysogenum (PC), Aspergillus niger (AN), Aspergillus terreus (AT), and Aspergillus
Zhaobin Mu et al.
Analytical and bioanalytical chemistry, 408(5), 1515-1522 (2015-12-31)
A multi-residue method based on modified QuEChERS (quick, easy, cheap, effective, rugged, and safe) sample preparation, followed by liquid chromatography tandem mass spectrometry (LC-MS/MS), was developed and validated for the determination of three selected fungicides (propiconazole, pyraclostrobin, and isopyrazam) in
Joanna Kania et al.
Pesticide biochemistry and physiology, 148, 166-174 (2018-06-13)
Cultivation of oilseed rape requires application of specific fungicides. Besides their protective role, they can potentially influence the expression and activity of crucial enzymes in the plant. Among the large number of enzymes expressed in plants, aminopeptidases play a key

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service