Skip to Content
Merck
All Photos(1)

Key Documents

18795

Millipore

Brucella Agar

suitable for microbiology, NutriSelect® Basic

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171607
NACRES:
NA.74

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 13 g/L
casein peptone, 10 g/L
D(+)-glucose, 1 g/L
meat peptone, 10 g/L
sodium chloride, 5 g/L
yeast extract, 2 g/L

manufacturer/tradename

NutriSelect® Basic

technique(s)

microbiological culture: suitable

final pH

7.0±0.2 (25 °C)

application(s)

cleaning products
clinical testing
cosmetics
environmental
food and beverages
personal care
veterinary

microbiology

suitability

Haemophilus spp.
Pneumococcus spp.
nonselective for Brucella spp.
nonselective for Listeria spp.
nonselective for Neisseria spp.
nonselective for Streptococcus spp.

Application

For the isolation and cultivation of Brucella species, in particular the pathogens B. melitensis, B. abortus and B. suis, from clinical material and comestibles of animal origin.

Preparation Note

Dissolve 41 g in 1 litre distilled water and autoclave at 121°C for 15 minutes. Let cool to 45-50°C and add the supplement according the data sheet.

Supplements :
Brucella differential agar:
1 ml/l 2% basic fuchsin solution (Cat. No. 03968). or 2% solution from thionine acetate (Cat. No. 88930)

Brucella selective agar:
500 mg/l Bacitracin (Cat. No. 11702)
1 mg/l Polymyxin B sulfate (Cat. No. P4932)
100 mg/l Cycloheximide (Cat. No. 01810)
(and if required) 1.25 mg/l ethyl violet (Cat. No. 228842)

For isolation of Campylobacter species:
50 ml/l heat inactivated horse serum (Cat. No. H1138)
250mg/l Sodium pyruvate (Cat. No. P2256)
250mg/l Sodium metabisulfite (Cat. No. 31448)
250mg/l Ferrous sulfate (Cat. No. 31236)
50-100 ml/l defibrinated sheep blood

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service