Skip to Content
Merck
All Photos(1)

Documents

14510

Sigma-Aldrich

1,2-Bis(2-aminophenoxy)ethane-N,N,N′,N′-tetraacetic acid

≥96.0% (HPLC)

Synonym(s):

BAPTA, Ethylenedioxybis(o-phenylenenitrilo)tetraacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H24N2O10
CAS Number:
Molecular Weight:
476.43
Beilstein:
5192406
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥96.0% (HPLC)

form

powder

fluorescence

λex 251 nm; λem 363 nm
λex 274 nm; λem 363 nm (With Calcium)

storage temp.

2-8°C

SMILES string

OC(=O)CN(CC(O)=O)c1ccccc1OCCOc2ccccc2N(CC(O)=O)CC(O)=O

InChI

1S/C22H24N2O10/c25-19(26)11-23(12-20(27)28)15-5-1-3-7-17(15)33-9-10-34-18-8-4-2-6-16(18)24(13-21(29)30)14-22(31)32/h1-8H,9-14H2,(H,25,26)(H,27,28)(H,29,30)(H,31,32)

InChI key

FTEDXVNDVHYDQW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,2-Bis(2-aminophenoxy)ethane-N,N,N′,N′-tetraacetic acid is a highly selective calcium chelating reagent. Effect of microinjected calcium-loaded 1,2-bis(2-aminophenoxy)ethane-N,N,N′,N′-tetraacetic acid (CaBAPTA) on cell-to-cell diffusion of carboxyfluorescein has been studied in staminal hairs of Setcreasea purpurea. It is reported to be a metabolite of 1,2-bis(2-aminophenoxy)ethane-N,N,N′,N′-tetraacetic acid tetra(acetoxymethyl ester) (BAPTA-AM) in rat urine.
pH 8.0 Highly selective calcium chelating reagent. Suitable for the spectrophotometric monitoring of Ca2+ levels

Application

1,2-Bis(2-aminophenoxy)ethane-N,N,N′,N′′-tetraacetic acid was used for the spectrophotometric monitoring of Ca2+ levels. It may be used as chelator of cytosolic Ca²+ to investigate the effect of methoxychlor on cytosolic free Ca²+ concentrations and apoptosis in HA59T human hepatoma cells.
Highly selective calcium chelating reagent. Suitable for the spectrophotometric monitoring of Ca2+ levels

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B E Britigan et al.
Biochemical pharmacology, 55(3), 287-295 (1998-03-04)
A role for increases in intracellular calcium (Ca2+) has been suggested in the pathophysiology of various forms of oxidant-mediated cell injury. In recent studies, we found that iron bound to the Pseudomonas aeruginosa siderophore, pyochelin, augments oxidant-mediated endothelial cell injury
E B Tucker
Planta, 182(1), 34-38 (1990-08-01)
The effect of microinjected calcium-loaded 1,2-bis(2-aminophenoxy) ethane-N,N,N',N'-tetraacetic acid (CaBAPTA) on cell-to-cell diffusion of carboxyfluorescein (CF) was examined in staminal hairs of S. purpurea Boom. The CaBAPTA was microinjected into the cytoplasm of the staminal hairs either with CF or prior
Chi-Ting Horng et al.
The Chinese journal of physiology, 58(1), 1-8 (2015-02-18)
Methoxychlor, an organochlorine pesticide, is thought to be an endocrine disrupter that affects Ca²⁺ homeostasis and cell viability in different cell models. This study explored the action of methoxychlor on cytosolic free Ca²⁺ concentrations ([Ca²⁺]i) and apoptosis in HA59T human
Determination of 1,2-bis(2-aminophenoxy) ethane-N,N,N',N'-tetraacetic acid (BAPTA free acid) in rat plasma, urine and feces by liquid chromatography with UV and tandem mass spectrometric detection.
Feng Z and Wenying L.
Journal of Chromatography. B, Biomedical Applications, 878(30), 3052-3058 (2010)
Mayeul Collot et al.
Journal of the American Chemical Society, 134(36), 14923-14931 (2012-07-24)
We designed Calcium Rubies, a family of functionalizable BAPTA-based red-fluorescent calcium (Ca(2+)) indicators as new tools for biological Ca(2+) imaging. The specificity of this Ca(2+)-indicator family is its side arm, attached on the ethylene glycol bridge that allows coupling the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service