Skip to Content
Merck
All Photos(1)

Documents

04054

Supelco

Acesulfame K

for food analysis, ≥99.0%

Synonym(s):

6-Methyl-1,2,3-oxathiazin-4(3H)-one 2,2-dioxide potassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4KNO4S
CAS Number:
Molecular Weight:
201.24
Beilstein:
3637857
EC Number:
MDL number:
UNSPSC Code:
12164500
PubChem Substance ID:
NACRES:
NA.21

grade

for food analysis

Quality Level

Assay

≥99.0% (HPLC)
≥99.0%

sweetness

200 × sucrose

application(s)

food and beverages

SMILES string

[K+].CC1=CC(=O)[N-]S(=O)(=O)O1

InChI

1S/C4H5NO4S.K/c1-3-2-4(6)5-10(7,8)9-3;/h2H,1H3,(H,5,6);/q;+1/p-1

InChI key

WBZFUFAFFUEMEI-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

′New generation′, heat-stable sweetener that has not been suspected to cause cancer nor be genotoxic. Allelic variation of the Tas1r3 gene affects behavioral taste responses to this molecule, suggesting that it is a T1R3 receptor ligand.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei-Li Li et al.
Journal of molecular neuroscience : MN, 51(1), 225-236 (2013-04-23)
The sweet taste is of immense interest to scientists and has been intensively studied during the last two decades. However, the sweet preference modification and the related mechanisms are still unclear. In this study, we try to establish a mice
Mi-Sun Ha et al.
International journal of food sciences and nutrition, 64(6), 715-723 (2013-05-02)
Using a stepwise assessment of the exposure of Korean consumers to acesulfame K and sucralose, theoretical maximum daily intakes of the sweeteners were calculated using the Budget screening method, which resulted in values greater than the acceptable daily intakes (ADIs).
Francesco Di Salle et al.
Gastroenterology, 145(3), 537-539 (2013-05-30)
Little is known about how CO2 affects neural processing of taste. We used functional magnetic resonance imaging to investigate the effects of carbonation on brain processing of sweet stimuli, which has relevance to studies of food selection and satiety. The
In search of a role for carbonation: is this a good or bad taste?
Catia Sternini
Gastroenterology, 145(3), 500-503 (2013-07-31)
Pornrat Sinchaipanit et al.
Journal of the science of food and agriculture, 93(13), 3304-3311 (2013-04-16)
Different ratios of combined sweeteners were modified to produce an acceptable reduced-calorie carrot juice. Various hydrocolloids were investigated to improve juice cloud stability. Changes in juice quality attributes were analysed. A combination of the sweeteners aspartame (ASP), acesulfame potassium (ACE)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service