Skip to Content
Merck
All Photos(1)

Key Documents

34-0013

Sigma-Aldrich

Mineral oil

CP

Synonym(s):

White mineral oil 70

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12181504
grade:
CP
Pricing and availability is not currently available.

grade

CP

availability

available only in Japan

refractive index

n20/D 1.467 (lit.)

InChI key

AEOVEGJBKQQFOP-DDVLFWKVSA-L

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takeshi Itabashi et al.
Nature methods, 6(2), 167-172 (2009-01-20)
Accurate chromosome segregation during meiosis depends on the assembly of a microtubule-based spindle of proper shape and size. Current models for spindle-size control focus on reaction diffusion-based chemical regulation and balance in activities of motor proteins. Although several molecular perturbations
Angela Rouyar et al.
PloS one, 6(10), e26443-e26443 (2011-10-27)
In nature the aerial trace of pheromone used by male moths to find a female appears as a train of discontinuous pulses separated by gaps among a complex odorant background constituted of plant volatiles. We investigated the effect of such
Xuchu Ma et al.
Dalton transactions (Cambridge, England : 2003), (46)(46), 10250-10255 (2009-11-19)
Na(4)Si(4) and Na(4)Ge(4) are ideal chemical precursors for inorganic clathrate structures, clusters, and nanocrystals. The monoclinic Zintl phases, Na(4)Si(4) and Na(4)Ge(4), contain isolated homo-tetrahedranide [Si(4)](4-) and [Ge(4)](4-) clusters surrounded by alkali metal cations. In this study, a simple scalable route
Keke Shao et al.
PloS one, 6(9), e24910-e24910 (2011-09-29)
Aptamers are short RNA or DNA oligonucleotides which can bind with different targets. Typically, they are selected from a large number of random DNA sequence libraries. The main strategy to obtain aptamers is systematic evolution of ligands by exponential enrichment
Benjamin Grau et al.
Purinergic signalling, 4(4), 357-364 (2008-09-09)
The formation of appropriate connections between neurons and their specific targets is an essential step during development and repair of the nervous system. Growth cones are located at the leading edges of the growing neurites and respond to environmental cues

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service