Skip to Content
Merck
All Photos(1)

Key Documents

11-0130

Sigma-Aldrich

Ferroin indicator solution

Synonym(s):

1,10-Phenanthroline iron(II) sulfate complex, o-Phenanthroline ferrous sulfate complex

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[Fe(C12H8N2)3]SO4
CAS Number:
Molecular Weight:
692.52
MDL number:
UNSPSC Code:
41116100
PubChem Substance ID:

availability

available only in Japan

SMILES string

[Fe++].[O-]S([O-])(=O)=O.c1cnc2c(c1)ccc3cccnc23.c4cnc5c(c4)ccc6cccnc56.c7cnc8c(c7)ccc9cccnc89

InChI

1S/3C12H8N2.Fe.H2O4S/c3*1-3-9-5-6-10-4-2-8-14-12(10)11(9)13-7-1;;1-5(2,3)4/h3*1-8H;;(H2,1,2,3,4)/q;;;+2;/p-2

InChI key

CIWXFRVOSDNDJZ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Suitability

for titration

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V H Tomasi et al.
Biotechnic & histochemistry : official publication of the Biological Stain Commission, 78(5), 255-259 (2004-03-03)
We describe here a new method for specific staining of mast cells using ferroin. Different hamster tissues were fixed in 4% formalin and processed for paraffin embedding. Sections were stained with hematoxylin followed by ferroin acidified with 2.5 N sulfuric
Catalina Peralta et al.
The journal of physical chemistry. A, 110(44), 12145-12149 (2006-11-03)
The purpose of this research was to explore the unstirred, ferroin-catalyzed Belousov-Zhabotinsky (BZ) reaction as an experimental model for the response of excitable media to small perturbations (slightly larger than the threshold for excitations). Following Showalter et al. (Showalter, K.;
Jichang Wang et al.
The journal of physical chemistry. A, 109(7), 1374-1381 (2006-07-13)
The ferroin-catalyzed Belousov-Zhabotinsky (BZ) reaction was studied in a batch reactor under anaerobic conditions and was found to evolve through two separated regimes of complex oscillations. Significantly, the two bifurcation regimes exhibited qualitatively different dependence on compositions of the reaction
Luciano A Cardoso et al.
Comparative biochemistry and physiology. Part A, Molecular & integrative physiology, 151(3), 313-321 (2007-06-05)
Glutathione reductase (GR) carries out the enzymatic reduction of glutathione disulfide (GSSG) to its reduced form (GSH) at the expense of the reducing power of NADPH. Previous studies have shown that GR from several species is progressively inactivated in the
C Fernandez-Patron et al.
Circulation research, 87(8), 670-676 (2000-10-13)
Matrix metalloproteinase (MMP)-2 has been historically associated with the process of vascular remodeling through the cleavage of extracellular matrix proteins. However, we recently found that MMP-2 also cleaves the endothelium-derived peptide big endothelin-1, ET-1[1-38] and yields the novel vasoconstrictor ET-1[1-32].

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service