Skip to Content
Merck
All Photos(1)

Documents

07-5640

Sigma-Aldrich

Diphenyl ether

SAJ first grade, ≥98.0%

Synonym(s):

Diphenyl oxide, Phenyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2O
CAS Number:
Molecular Weight:
170.21
Beilstein:
1364620
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:

grade

SAJ first grade

vapor density

>5.86 (25 °C, vs air)

vapor pressure

<1 mmHg ( 20 °C)

Assay

≥98.0%

form

crystalline

autoignition temp.

1144 °F

expl. lim.

1.5 %

availability

available only in Japan

refractive index

n20/D 1.579 (lit.)

bp

259 °C (lit.)

mp

25-27 °C (lit.)

density

1.073 g/mL at 25 °C (lit.)

SMILES string

O(c1ccccc1)c2ccccc2

InChI

1S/C12H10O/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12/h1-10H

InChI key

USIUVYZYUHIAEV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

239.0 °F - closed cup

Flash Point(C)

115 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kostantinos Benekos et al.
Journal of biotechnology, 150(1), 195-201 (2010-07-20)
Plant glutathione transferases (GSTs) superfamily consists of multifunctional enzymes and forms a major part of the plants herbicide detoxification enzyme network. The tau class GST isoenzyme GmGSTU4 from soybean, exhibits catalytic activity towards the diphenyl ether herbicide fluorodifen and is
Ariana Beste et al.
The Journal of organic chemistry, 74(7), 2837-2841 (2009-03-06)
Lignin is an abundant natural resource that is a potential source of valuable chemicals. Improved understanding of the pyrolysis of lignin occurs through the study of model compounds for which phenethyl phenyl ether (PhCH(2)CH(2)OPh, PPE) is the simplest example representing
Ge-Fei Hao et al.
Journal of computer-aided molecular design, 25(3), 213-222 (2011-01-25)
Protoporphyrinogen oxidase (PPO, EC 1.3.3.4), which has been identified as a significant target for a great family of herbicides with diverse chemical structures, is the last common enzyme responsible for the seventh step in the biosynthetic pathway to heme and
Tony Ly et al.
Analytical chemistry, 80(13), 5059-5064 (2008-05-24)
Selective noncovalent adduct protein probing (SNAPP) mass spectrometry was recently developed to study solution-phase conformations of proteins by exploiting the specific affinity between 18-crown-6 ether (18C6) and lysine side chains. To obtain more detailed information about protein tertiary structure, a
Bruno F B Silva et al.
Langmuir : the ACS journal of surfaces and colloids, 31(14), 4361-4371 (2014-11-15)
In this work we investigate the interplay between flow and boundary condition effects on the orientation field of a thermotropic nematic liquid crystal under flow and confinement in a microfluidic device. Two types of experiments were performed using synchrotron small-angle

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service