Skip to Content
Merck
All Photos(1)

Key Documents

07-2670

Sigma-Aldrich

Diethanolamine

SAJ first grade, ≥98.0%

Synonym(s):

2,2′-Iminodiethanol, Bis(2-hydroxyethyl)amine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HN(CH2CH2OH)2
CAS Number:
Molecular Weight:
105.14
Beilstein:
605315
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

grade

SAJ first grade

vapor density

3.6 (vs air)

vapor pressure

<0.98 atm ( 100 °C)

Assay

≥98.0%

form

liquid

autoignition temp.

689 °F

expl. lim.

10.6 %

availability

available only in Japan

refractive index

n20/D 1.477 (lit.)

pH

11.0-12 (25 °C, 105 g/L)

bp

217 °C/150 mmHg (lit.)

mp

28 °C (lit.)

density

1.097 g/mL at 25 °C (lit.)

SMILES string

OCCNCCO

InChI

1S/C4H11NO2/c6-3-1-5-2-4-7/h5-7H,1-4H2

InChI key

ZBCBWPMODOFKDW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 Oral

Target Organs

Kidney,Liver,Blood

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

280.4 °F - closed cup

Flash Point(C)

138 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

William Conway et al.
Environmental science & technology, 46(13), 7422-7429 (2012-05-25)
The kinetics of the fast reversible carbamate formation reaction of CO(2)(aq) with a series of substituted cyclic secondary amines as well as the noncyclic secondary amine diethanolamine (DEA) has been investigated using the stopped-flow spectrophotometric technique at 25.0 °C. The
Siou-Yin Chen et al.
Inorganic chemistry, 51(8), 4448-4457 (2012-04-05)
The synthesis, X-ray crystallography, magnetic properties, and high-field electron paramagnetic resonance (HFEPR) of a new heptanuclear manganese complex [Mn(7)(heamp)(6)](ClO(4))(2)·4CH(2)Cl(2)·H(2)O (complex 2), in which heampH(3) is 2-[N,N-di(2-hydroxyethyl)aminomethyl]phenol (compound 1), is reported. Complex 2 has a hexagonal, disk-shaped topology and contains six
Honghui Yu et al.
Journal of immunology (Baltimore, Md. : 1950), 193(6), 2902-2910 (2014-08-15)
Anti-D can prevent immunization to the RhD Ag on RBCs, a phenomenon commonly termed Ab-mediated immune suppression (AMIS). The most accepted theory to explain this effect has been the rapid clearance of RBCs. In mouse models using SRBC, these xenogeneic
L Jiménez et al.
Bioresource technology, 100(3), 1478-1481 (2008-09-24)
The AOpAZRP bleaching sequence (A is an acid treatment, Op an oxygen and peroxide stage, Z an ozone stage, R a reductive treatment and P a peroxide stage) have been applied to oil palm empty fruit bunches (EFB) soda-anthraquinone and
Bjørn Henrik Hansen et al.
Aquatic toxicology (Amsterdam, Netherlands), 99(2), 212-222 (2010-06-12)
Alkanolamines are surface-active chemicals used in a wide range of industrial, agricultural and pharmaceutical applications and products. Of particular interest is the use of alkanolamines such as diethanolamine (DEA) in the removal of CO(2) from natural gas and for CO(2)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service