Skip to Content
Merck
All Photos(1)

Key Documents

03-0700

Sigma-Aldrich

Bentonite

CP

Synonym(s):

Montmorillonite

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
23201100

grade

CP

form

granular

availability

available only in Japan

SMILES string

O.O=[Si]=O.O=[Si]=O.O=[Si]=O.O=[Si]=O.O=[Al]O[Al]=O

InChI

1S/2Al.4O2Si.H2O.3O/c;;4*1-3-2;;;;/h;;;;;;1H2;;;

InChI key

KARVSHNNUWMXFO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hanzhong Jia et al.
Environmental technology, 34(1-4), 25-33 (2013-03-28)
To mitigate the aggregation and enhance the reactivity of nanosized zero-valent iron (nZVI), montmorillonite is employed as a template-supporting matrix to prepare nZVI through two different pathways: heterogeneous nucleation and homogeneous nucleation processes. Dispersed sub-nanosized ZVI clusters with an average
D Saravanan et al.
International journal of biological macromolecules, 53, 67-71 (2012-11-15)
Contamination of water by toxic heavy metals due to urbanization is a world-wide environmental problem, which changes chemical and biological properties of both surface and ground water. The heavy metals render the water unsuitable for drinking and are also highly
Abdallah M Shanableh et al.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering, 48(2), 223-231 (2012-10-10)
This study was part of a larger effort that involves evaluating alternatives to upgrading secondary treatment systems in the United Arab Emirates for the removal of nutrients. In this study, six modified bentonite (BNT) phosphate adsorbents were prepared using solutions
C D Wu et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 67(1), 152-158 (2012-11-07)
The single-solute and bisolute sorption behaviour of phenol and trichloroethylene, two organic compounds with different structures, onto cetyltrimethylammonium bromide (CTAB)-montmorillonite was studied. The monolayer Langmuir model (MLM) and empirical Freundlich model (EFM) were applied to the single-solute sorption of phenol
T Corrales et al.
Biomacromolecules, 13(12), 4247-4256 (2012-11-17)
A triblock copolymer based on poly(ε-caprolactone) (PCL) and 2-(N,N-diethylamino)ethyl methacrylate (DEAEMA)/2-(methyl-7-nitrobenzofurazan)amino ethyl acrylate (NBD-NAcri), was synthesized via atom transfer radical polymerization (ATRP). The corresponding chlorohydrated copolymer, named as PCL-b-DEAEMA, was prepared and anchored via cationic exchange on montmorillonite (MMT) surface.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service