Skip to Content
Merck
All Photos(1)

Key Documents

01-3540

Sigma-Aldrich

6-Aminocaproic acid

SAJ special grade, ≥99.0%

Synonym(s):

ε-Aminocaproic acid, 6-Aminohexanoic acid, EACA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2N(CH2)5CO2H
CAS Number:
Molecular Weight:
131.17
Beilstein:
906872
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

grade

SAJ special grade

Assay

≥99.0%

form

solid

availability

available only in Japan

mp

207 °C
207-209 °C (dec.) (lit.)

solubility

water: soluble

application(s)

peptide synthesis

SMILES string

NCCCCCC(O)=O

InChI

1S/C6H13NO2/c7-5-3-1-2-4-6(8)9/h1-5,7H2,(H,8,9)

InChI key

SLXKOJJOQWFEFD-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Lysine analog. Promotes rapid dissociation of plasmin, thereby inhibiting the activation of plasminogen and subsequent fibrinolysis. Reported to inhibit plasminogen binding to activated platelets. An early report indicated that it inhibits the activation of the first component of the complement system. Binds and inactivates Carboxypeptidase B.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E M Van Hoeyveld et al.
The Journal of allergy and clinical immunology, 76(4), 543-550 (1985-10-01)
The effect of epsilon-aminocaproic acid (EACA) on the degradation of an aqueous Lolium perenne extract was studied by intracutaneous tests and by RAST inhibition. Extracts for skin testing stored at 4 degrees C for 12 months and at 37 degrees
P A Stricker et al.
British journal of anaesthesia, 110(5), 788-799 (2013-01-29)
Understanding the clinical pharmacology of the antifibrinolytic epsilon-aminocaproic acid (EACA) is necessary for rational drug administration in children. The aim of this study is to determine the pharmacokinetics (PKs) of EACA in infants aged 6-24 months undergoing craniofacial reconstruction surgery.
Moumita Ghosh et al.
Langmuir : the ACS journal of surfaces and colloids, 28(3), 1715-1724 (2011-12-29)
The present work reports covalent functionalization of single-walled carbon nanotubes (f-SWNTs) to introduce hydrophilicity to the otherwise amphiphobic nanotubes. The charge and spacer length of the functional moiety were varied by using quaternized ethylene diamine, 6-aminocaproate, quaternized (ethylenedioxy)bis(ethylamine), and a
Illa Ramakanth et al.
The journal of physical chemistry. B, 116(9), 2722-2729 (2012-02-09)
A two-component gel resulting from the amphiphilic cationic surfactant cetylpyridinium chloride (CPC) in the presence of a structure-forming bola-amphiphilic additive 6-aminocaproic acid (6-ACA) was realized and investigated. At a critical 6 wt % of 1:1 CPC:6-ACA, the yellow colored gel
Brian Hutton et al.
BMJ (Clinical research ed.), 345, e5798-e5798 (2012-09-13)
To estimate the relative risks of death, myocardial infarction, stroke, and renal failure or dysfunction between antifibrinolytics and no treatment following the suspension of aprotinin from the market in 2008 for safety reasons and its recent reintroduction in Europe and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service