Skip to Content
Merck
All Photos(1)

Key Documents

TEFG02525

Millipore

Steritest® vent needles

suitable for sterility testing, for glass vials with rubber septa and rigid plastic vials., PTFE (hydrophobic membrane filter)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151818
eCl@ss:
32014001
NACRES:
NB.24

material

PTFE (hydrophobic membrane filter)
PVC device
stainless steel (for needle)

Quality Level

Agency

EP (2.6.1)
JP (4.06)
USP 71

sterility

sterile; ethylene oxide treated

packaging

pkg of 25 units

manufacturer/tradename

Steritest®

filter size

24.7 mm

pore size

0.2 μm pore size

input

sample type pharmaceutical(s)

application(s)

pharmaceutical
sterility testing

compatibility

for use with Steritest® Symbio FLEX Pump Kit, 2 media (SYMBFLE01)
for use with Steritest® Symbio ISL Pump Kit, 2 media (SYMBISL01)
for use with Steritest® Symbio LFH Pump Kit (SYMBLFH01)

shipped in

ambient

General description

Steritest® vent needle is an accessory of the Steritest® NEO devices, used for liquid transfer and rinsing during sterility testing. It is a single needle vented with PTFE (hydrophobic) membrane.

Application

The Steritest® vent needles are used for venting glass vials with rubber septa and rigid plastic vials. They can also be used for rinsing of medical devices, vial-to-vial aseptic transfer of large volume parenteral, sterility and growth promotion qualification of media batches, or for venting of media bottles during the direct inoculation method.

Legal Information

STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service