Skip to Content
Merck
All Photos(1)

Key Documents

SCR032

Sigma-Aldrich

Mouse Cortical Neural Stem Cell Expansion Kit

The Mouse Cortical Neural Stem Cell Expansion Kit provides a multi-component system for the culture & analysis of neural stem cells & their differentiated progenies.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
NACRES:
NA.76

Quality Level

product line

Chemicon®

manufacturer/tradename

Chemicon®

technique(s)

cell culture | stem cell: suitable
immunocytochemistry: suitable

input

sample type neural stem cell(s)

General description

The Mouse Cortical Neural Stem Cell Expansion Kit provides a multi-component system for the culture and analysis of neural stem cells and their differentiated progenies. This system includes mouse cortical neural stem cells, isolated from the cortices of embryonic day 15-18 (E15-E18) C57/BL6 mice, neural stem cell expansion medium, and a panel of antibodies for immunocytochemical staining of neural stem/progenitor cells (Nestin and Sox2) and differentiated neural phenotypes (Map2ab for neurons, GFAP for astrocytes, and O1 for oligodendrocytes).

Application

The Mouse Cortical Neural Stem Cell Expansion Kit provides a multi-component system for the culture & analysis of neural stem cells & their differentiated progenies.

Components

  • Cryopreserved Mouse Cortical Neural Stem Cells (Cat.# SCR029)
  • Mouse Neural Stem Cell Expansion Medium (Cat.# SCM008)
  • Neural Stem Cell Marker Characterization Kit (Cat.# SCR019)

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service