Skip to Content
Merck
All Photos(2)

Documents

SCGPCAPRE

Millipore

Millipore® Stericap Plus Vacuum Filter

pore size 0.22 μm, polyethersulfone membrane, pack of 10 ea

Synonym(s):

SteriCap PLUS, universal bottle top sterile filter, universal bottle top vacuum filter, universal bottle top vacuum filtering unit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104914
eCl@ss:
32031623
NACRES:
NB.24

material

polyethersulfone membrane
polyethylene
polypropylene
polystyrene

Quality Level

sterility

sterile

feature

holdup volume 2.7 mL
hydrophilic

packaging

pack of 10 ea

parameter

45 °C max. temp.

technique(s)

sterile filtration: suitable

H

76 mm

diam.

10 cm

filter diam.

73 mm

filtration area

40 cm2

matrix

Express PLUS

pore size

0.22 μm pore size

fitting

inlet hose barb
outlet bottle top (Universal)

shipped in

ambient

General description

The Stericap PLUS Universal Bottle-Top Filter Device with Millipore Express PLUS membrane provides fast vacuum filtration and low protein binding without the need for a prefilter in most cases. It filters quickly, without clogging, even viscous solutions typical of serum-containing media. This non-cytotoxic, non-pyrogenic device can be used to fill any vacuum-rated bottle or other container with an inner neck diameter of 20 to 67 mm. This allows maximum versatility, capacity, speed, and flowthrough efficiency when you need to filter larger volumes into containers with openings of various sizes.

Features & Benefits:
- Millipore Express PLUS membrane for fastest flow rates and low protein binding

- Fits any vacuum-rated bottle from 20 to 67 mm in diameter for maximum flexibility

- High volume capacity for larger and less frequent preparation steps

- Easy flow control button so the filter can be easily transferred from bottle to bottle without turning off the vacuum each time

- Free of cytotoxic and pyrogenic contaminants for robust cell culture and sensitive downstream applications

Applications:
Sterilization of Aqueous Solutions, Media Preparation, Buffer Preparation

Application

For research use only.

Principle

Vacuum

Physical form

Color Code: Clear

Preparation Note

Sterilization Method
Gamma irradiation

Other Notes

Directions for Use

  • Organism Retention: Microorganisms
  • Mode of Action: Filtering
  • Application: Sterile filtration
  • Intended Use: Sterile filtration of laboratory reagents, buffers, and media.
  • Instructions for Use: Please refer to the instructions for use section of Stericup® PLUS user guide
  • Storage Statement: Store in dry location
  • Disposal Statement: Dispose of in accordance with applicable federal, state and local regulations.

Legal Information

EXPRESS PLUS is a trademark of Merck KGaA, Darmstadt, Germany
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany
STERICAP is a trademark of Merck KGaA, Darmstadt, Germany
Stericup is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service